Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
- Catalog No:
- AATB100521U0
- AATB100521U1
- AATB100521U2
- Antigen symbol:
- CD5
- CD5
- CD5
- Antigen name:
- CD5 antigen
- CD5 antigen
- CD5 antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- UCHT2
- UCHT2
- UCHT2
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
- 20
- Pack typePk
Specifications
Antigen symbolCD5Antigen nameCD5 antigenEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationPerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®CloneUCHT2ReactivityHumanHostMouseIsotypeImmunoFluorescenceYesFlow cytometryYesPurificationAffinity chromatographyCross adsorptionNoStorage temperature2...8 °C with minimized light exposure. Do not freeze.Shipping temperature20- AATB100521U1100521U1Vial100 Tests1 * 100 Tests4 160 zł
Specifications
Antigen symbolCD5Antigen nameCD5 antigenEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationPerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®CloneUCHT2ReactivityHumanHostMouseIsotypeImmunoFluorescenceYesFlow cytometryYesPurificationAffinity chromatographyCross adsorptionNoStorage temperature2...8 °C with minimized light exposure. Do not freeze.Shipping temperature20 - AATB100521U2100521U2Vial500 Tests1 * 500 Tests14 090 zł
Specifications
Antigen symbolCD5Antigen nameCD5 antigenEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationPerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®CloneUCHT2ReactivityHumanHostMouseIsotypeImmunoFluorescenceYesFlow cytometryYesPurificationAffinity chromatographyCross adsorptionNoStorage temperature2...8 °C with minimized light exposure. Do not freeze.Shipping temperature20