About this item
Discovery® BIO Wide Pore reversed-phase columns satisfy the need for efficiency, selectivity, scalability, and reproducibility for RP HPLC analyses of proteins, peptides, and small biomolecules.
- Better protein and peptide resolution compared to leading RP-HPLC FPP columns
- High efficiency for peptide mapping
- Low-bleed, inert surface chemistry make them ideal for LC-MS applications
- Reliable reproducibility run -to-run, column-to-column, batch-to-batch
- Separation are completely scalable from analytical to preparative dimensions
- Surface coverage: 3,6 µmol/m²
Discovery® BIO Wide Pore HPLC columns are packed with C18 ligands bonded to 3, 5, or 10 µm spherical, high purity silica particles containing 300 Å pores.
The low-bleed character and excellent peak shape without TFA in the mobile phase makes these columns ideal for proteomics and other LC-MS applications and preparative purification.
Specifications
- Catalog No:
- 567271-U
- 567270-U
- 568272-U
- 568271-U
- 567208-U
- 567207-U
- 568230-U
- 568223-U
- 567200-U
- 568213-U
- 568203-U
- 567209-U
- 567205-U
- 568273-U
- 567204-U
- 568222-U
- 567212-U
- 567202-U
- 567211-U
- 567201-U
- Application:
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Peptide mapping/proteolytic digests/small biomolecules
- Carbon load:
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- 9,2%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- Temperature limit:
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Shipping conditions:
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
Specifications
Analytical columns, particle size 3 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC columnHPLC column567200-U567200-UAnalyticalC18e300 Å3 µm2,1 mmLog in for availability1 * 1 Szt.3 130 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567205-U567205-UAnalyticalC18e300 Å3 µm4,6 mmLog in for availability1 * 1 Szt.4 920 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567204-U567204-UAnalyticalC18e300 Å3 µm4,6 mmLog in for availability1 * 1 Szt.4 480 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567202-U567202-UAnalyticalC18e300 Å3 µm2,1 mmLog in for availability1 * 1 Szt.4 920 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567201-U567201-UAnalyticalC18e300 Å3 µm2,1 mmLog in for availability1 * 1 Szt.4 480 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water
Guard cartridges, particle size 3 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC guard cartridge (pack of 2 each)HPLC guard cartridge (pack of 2 each)567270-U567270-UGuard cartridgeC18e300 Å3 µm2,1 mmLog in for availability1 * 1 Szt.1 270 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC guard cartridge kitHPLC guard cartridge kit567271-U567271-UGuard cartridgeC18e300 Å3 µm2,1 mmLog in for availability1 * 1 Szt.1 140 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water
Analytical columns, particle size 5 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC columnHPLC column568203-U568203-UAnalyticalC18e300 Å5 µm2,1 mmLog in for availability1 * 1 Szt.4 480 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column568213-U568213-UAnalyticalC18e300 Å5 µm4,0 mmLog in for availability1 * 1 Szt.4 480 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column568223-U568223-UAnalyticalC18e300 Å5 µm4,6 mmLog in for availability1 * 1 Szt.4 480 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column568230-U568230-UAnalyticalC18e300 Å5 µm10,0 mmLog in for availability1 * 1 Szt.9 860 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column568222-U568222-UAnalyticalC18e300 Å5 µm4,6 mmLog in for availability1 * 1 Szt.4 110 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water
Guard cartridges, particle size 5 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC guard cartridge (1 pack of 2 each; 2 guard columns)HPLC guard cartridge (1 pack of 2 each; 2 guard columns)568272-U568272-UGuard cartridgeC18e300 Å5 µm4,0 mmLog in for availability1 * 1 Szt.1 270 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC guard cartridge (kit contains 1 guard column and a stand-alonge holder, a piece of tubing and 2 nuts and ferrules)HPLC guard cartridge (kit contains 1 guard column and a stand-alonge holder, a piece of tubing and 2 nuts and ferrules)568271-U568271-UGuard cartridgeC18e300 Å5 µm2,1 mmLog in for availability1 * 1 Szt.1 140 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC guard cartridge (kit contains 1 guard column and a stand-alonge holder, a piece of tubing and 2 nuts and ferrules)HPLC guard cartridge (kit contains 1 guard column and a stand-alonge holder, a piece of tubing and 2 nuts and ferrules)568273-U568273-UGuard cartridgeC18e300 Å5 µm4,0 mmLog in for availability1 * 1 Szt.1 270 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water
Analytical columns, particle size 10 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC columnSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567208-U567208-UAnalyticalC18e300 Å10 µm10,0 mmLog in for availability1 * 1 Szt.7 490 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnHPLC column567209-U567209-UAnalyticalC18e300 Å10 µm10,0 mmLog in for availability1 * 1 Szt.9 170 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water
Preparative columns, particle size 10 µm
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.AvailabilityPrice
- HPLC columnHPLC column567212-U567212-UPreparativeC18e300 Å10 µm21,2 mmLog in for availability1 * 1 Szt.17 020 złSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water - HPLC columnSpecifications:
More
Specifications
Environmentally PreferableApplicationPeptide mapping/proteolytic digests/small biomoleculesCarbon load9,2%EndcappedYesMax. pressure400 barpH range2 - 8Surface area100 m²/gTemperature limit0...70 °CUSP codeL1Shipping conditionsAcetonitril/water