Order Entry
Fluorescence Cuvettes for Spectroscopy
Fluorescence Cuvettes for Spectroscopy
Supplier:  HELLMA
New Product
Certificates
Fluorescence Cuvettes for Spectroscopy
Supplier:  HELLMA
New Product
Product Information
  • Catalog #
    Supplier #
    Type
    Base thickness
    Volume
    Path length
    Window material
    Availability
    Price
  • Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×40 mm, inside width: 2 mm
    Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×40 mm, inside width: 2 mm
    HELL115-F-10-K-40
    115-F-10-K-40
    Micro
    2 mm
    400 µl
    10×2 mm
    115F-QS
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT
    Supplier Article Number: 115-F-10-K-40
    Local Article Number: HELL115-F-10-K-40
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×46 mm, inside width: 4 mm
    Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×46 mm, inside width: 4 mm
    HELL114F-10-K-40
    114F-10-K-40
    Semi-micro
    1,25 mm
    1400 µl
    10×4 mm
    114F-QS
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT
    Supplier Article Number: 114F-10-K-40
    Local Article Number: HELL114F-10-K-40
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×46 mm, inside width: 10 mm, 4 windows
    Fluorescence cells, with PTFE lid or stopper, W×D×H: 12,5×12,5×46 mm, inside width: 10 mm, 4 windows
    HELL111-10-P-40
    111-10-P-40
    Macro
    1,25 mm
    3500 µl
    10×10 mm
    111-QS
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 111-10-P-40
    Local Article Number: HELL111-10-P-40
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • Fluorescence cells, macro, semi-micro, for anaerobic applications, W×D×H: 12,5×12,5×56 mm, inside width: 10 mm, 4 windows
    Fluorescence cells, macro, semi-micro, for anaerobic applications, W×D×H: 12,5×12,5×56 mm, inside width: 10 mm, 4 windows
    HELL117-100F10-K-4
    117-100F10-K-4
    Sealable
    1,25 mm
    3500 µl
    10×10 mm
    117.100F-QS
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT
    Supplier Article Number: 117-100F10-K-4
    Local Article Number: HELL117-100F10-K-4
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • Fluorescence cells, macro, semi-micro, with PTFE lid or stopper, W×D×H: 12,5×12,5×49,5 mm, inside width: 10 mm, 4 windows
    Fluorescence cells, macro, semi-micro, with PTFE lid or stopper, W×D×H: 12,5×12,5×49,5 mm, inside width: 10 mm, 4 windows
    HELL119F-10-K-40
    119F-10-K-40
    Magnetic stirrers
    5 mm
    3500 µl
    10×10 mm
    119.000F-QS
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT
    Supplier Article Number: 119F-10-K-40
    Local Article Number: HELL119F-10-K-40
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product