Adapters, delivery, bent
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Product Information
- Catalog #Supplier #Socket sizeCone sizePkAvailabilityPrice
- LENZ052650010526500114/23 NS14/23 NS11 * 1 SZT109 złSpecifications:
Specifications
Environmentally Preferable - 201-31090526500219/26 NS19/26 NS11 * 1 SZT121 złSpecifications:
Specifications
Environmentally Preferable - 201-31100526500429/32 NS29/32 NS11 * 1 SZT148 złSpecifications:
Specifications
Environmentally Preferable - LENZ052650030526500324/29 NS24/29 NS11 * 1 SZT188 złSpecifications:
Specifications
Environmentally Preferable