Order Entry
Beakers, low form
Beakers, low form
Catalog # 213-0185
Supplier:  KARTELL
Supplier #: 1541
undefined
Beakers, low form
Catalog # 213-0185
Supplier:  KARTELL
Supplier #: 1541
Price
35,50 zł
Log in for availability
Select an Option
34 mm
Select an Option
1 ml
Select an Option
25 ml
Select an Option
49 mm
Select an Option
01
Select an Option
1
Unit of Measure 1 * 1 SZT
About this item
PMP (TPX), crystal clear.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read embossed graduations.
ISO 7056
Certifications: ISO 7056
Product Information
  • Catalog #
    Supplier #
    Capacity
    Height
    Ext.Ø
    Graduation Interval
    Pk
    Availability
    Price
  • 213-0185
    1541
    25 ml
    49 mm
    34 mm
    1 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 35,50 zł
    35,50 zł
    1 * 1 SZT
       35,50 zł
    Supplier Article Number: 1541
    Local Article Number: KART1541
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0186
    1542
    50 ml
    60 mm
    42 mm
    2 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 42,50 zł
    42,50 zł
    1 * 1 SZT
       42,50 zł
    Supplier Article Number: 1542
    Local Article Number: KART1542
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0187
    1543
    100 ml
    72 mm
    52 mm
    5 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 43,50 zł
    43,50 zł
    1 * 1 SZT
       43,50 zł
    Supplier Article Number: 1543
    Local Article Number: KART1543
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0188
    1545
    250 ml
    95 mm
    71 mm
    10 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 54,50 zł
    54,50 zł
    1 * 1 SZT
       54,50 zł
    Supplier Article Number: 1545
    Local Article Number: KART1545
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0189
    1546
    500 ml
    119 mm
    88 mm
    10 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 89 zł
    89 zł
    1 * 1 SZT
       89 zł
    Supplier Article Number: 1546
    Local Article Number: KART1546
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0190
    1548
    1000 ml
    146 mm
    110 mm
    20 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 117 zł
    117 zł
    1 * 1 SZT
       117 zł
    Supplier Article Number: 1548
    Local Article Number: KART1548
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0191
    1549
    2000 ml
    184 mm
    133 mm
    50 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 177 zł
    177 zł
    1 * 1 SZT
       177 zł
    Supplier Article Number: 1549
    Local Article Number: KART1549
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0183
    1330
    3000 ml
    200 mm
    159 mm
    500 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 262 zł
    262 zł
    1 * 1 SZT
       262 zł
    Supplier Article Number: 1330
    Local Article Number: KART1330
    Specifications:
    Specifications
    Environmentally Preferable
  • 213-0184
    1331
    5000 ml
    228 mm
    190 mm
    500 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 430 zł
    430 zł
    1 * 1 SZT
       430 zł
    Supplier Article Number: 1331
    Local Article Number: KART1331
    Specifications:
    Specifications
    Environmentally Preferable