Order Entry
HPLC/UHPLC columns, Kromasil® Classic, 100 Å
  HISMM05CMA25
 :  HICHROM
HPLC/UHPLC columns, Kromasil® Classic, 100 Å
  HISMM05CMA25
 :  HICHROM
2 700 zł
  1 * 1 SZT
Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    100 Å
  • Length:
    250 mm
  • Seq:
    26
  • Phase:
    C8
  • Pk:
    1
Specifications
Bulk media
  •  
    Seq
    Description
    Particle Size
    Pore size
    Volume
    Pk
  • 04
    Kromasil C18(w)
    10 µm
    100 Å
    2 kg
    1
    1 * 1 SZT
    1 * 1 SZT - 66 610 zł
    66 610 zł
  •  
    Seq
    Description
    Particle Size
    Pore size
    Volume
    Pk
  • 02
    Kromasil C18(w)
    10 µm
    100 Å
    500 g
    1
    1 * 1 SZT
    1 * 1 SZT - 20 240 zł
    20 240 zł
  •  
    Seq
    Description
    Particle Size
    Pore size
    Volume
    Pk
  • 01
    Kromasil C18(w)
    10 µm
    100 Å
    200 g
    1
    1 * 1 SZT
    1 * 1 SZT - 9 340 zł
    9 340 zł
  •  
    Seq
    Description
    Particle Size
    Pore size
    Volume
    Pk
  • 03
    Kromasil C18(w)
    10 µm
    100 Å
    1 kg
    1
    1 * 1 SZT
    1 * 1 SZT - 35 980 zł
    35 980 zł