VWR®, Crucibles, Porcelain
About this item
Product Information
- Catalog #TypeCapacityØ×HPkAvailabilityPrice
- 459-0194Low form5 ml30×19 mm51 * 5 SZT144 złLocal Article Number: VWRI459-0194Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0195Low form10 ml35×22 mm51 * 5 SZT151 złLocal Article Number: VWRI459-0195Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0196Low form17 ml40×25 mm51 * 5 SZT163 złLocal Article Number: VWRI459-0196Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0197Low form21 ml45×28 mm51 * 5 SZT193 złLocal Article Number: VWRI459-0197Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0198Low form34 ml50×32 mm51 * 5 SZT269 złLocal Article Number: VWRI459-0198Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0199Low form62 ml60×38 mm51 * 5 SZT295 złLocal Article Number: VWRI459-0199Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0200Low form91 ml70×44 mm51 * 5 SZT395 złLocal Article Number: VWRI459-0200Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0201Medium form10 ml30×25 mm51 * 5 SZT146 złLocal Article Number: VWRI459-0201Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0202Medium form12 ml35×28 mm51 * 5 SZT151 złLocal Article Number: VWRI459-0202Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0203Medium form20 ml40×32 mm51 * 5 SZT166 złLocal Article Number: VWRI459-0203Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0204Medium form30 ml45×36 mm51 * 5 SZT199 złLocal Article Number: VWRI459-0204Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0205Medium form45 ml50×40 mm51 * 5 SZT247 złLocal Article Number: VWRI459-0205Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0206Medium form80 ml60×48 mm51 * 5 SZT306 złLocal Article Number: VWRI459-0206Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0207Medium form120 ml70×56 mm51 * 5 SZT402 złLocal Article Number: VWRI459-0207Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0208Medium form200 ml80×62 mm51 * 5 SZT546 złLocal Article Number: VWRI459-0208Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0209Tall form15 ml30×38 mm51 * 5 SZT173 złLocal Article Number: VWRI459-0209Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0210Tall form26 ml35×44 mm51 * 5 SZT188 złLocal Article Number: VWRI459-0210Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0211Tall form35 ml40×50 mm51 * 5 SZT203 złLocal Article Number: VWRI459-0211Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0212Tall form50 ml45×56 mm51 * 5 SZT244 złLocal Article Number: VWRI459-0212Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0213Tall form72 ml50×62 mm51 * 5 SZT332 złLocal Article Number: VWRI459-0213Specifications:
Accessories
Specifications
Environmentally Preferable - 459-0214Tall form130 ml60×75 mm51 * 5 SZT457 złLocal Article Number: VWRI459-0214Specifications:
Accessories
Specifications
Environmentally Preferable
Lids for crucibles
Product Information
- Catalog #SeqFor ØPkPriceAvailabilityPrice
- Local Article Number: VWRI459-0221