Drukuj
Naczynia hodowlane do zapłodnienia, Falcon®
Dostawca: Corning
Falcon®
734-2406EA 510 PLN
734-2406 734-0398 734-0068
Naczynia hodowlane do zapłodnienia, Falcon®
Szalki do hodowli Szalki do hodowli komórkowych
PS, sterile, with lid. Pre-tested and certified plasticware for in vitro fertilisation (IVF), designed for consistency and ease of use. These dishes have perfectly flat, optically clear surfaces for optimum manipulation and observation of ova and embryos. Lids are designed for aseptic manipulation and consistent venting to maintain humidification.
  • Non-embryotoxic, non-pyrogenic and non-cytotoxic
  • Tissue culture treated for a consistent hydrophilic surface
  • Sterilised by gamma irradiation
  • Packaged in peel-open, medical-style packaging
  • Multi-unit bags have reseal tabs
They have flat, optically clear surfaces for optimum manipulation and observation. Lids are designed for aseptic manipulation and consistent venting to maintain humidification. Sterilized by gamma irradiation. Nonembryotoxic (the lids of the Falcon IVF Products are not certified nonembryotoxic), nonpyrogenic, and noncytotoxic.
Informacje do zamówienia: Dishes are supplied with lids and packaged in resealable bags.