Chcesz zalogować się jeszcze raz?
Szalki Petriego, Gosselin™
Dostawca: Corning®
Gosselin
Contact plates used in hygiene control within the pharmaceutical or food processing industry, as well as in hospital environments. Manufactured in a Class 100 environment with cleanroom packaging. Available aseptic or sterile (SAL 10-6).
  • Moulded grid on the bottom makes it possible to identify contamination per cm²
  • Grip lid ensures safety during transport
  • Domed base guarantees a saving in agar whilst maintaining safe sampling
  • Stable stacking
ZAMÓW
Rozwiń / Zwiń
Wersja Ø Wysokość Sterylność Typ pakowania Ilość Numer katalogowy VWR Ilość
Domed, with 4 vents 65 mm 14,5 mm - 20/bag 720 391-0485
Domed, with 4 vents 65 mm 14,5 mm + 20/bag 720 391-0484
Domed, without vents 65 mm 14,5 mm - 20/bag 720 391-0483
With 3 vents 65 mm 14,5 mm - 20/bag 720 391-2064
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość