Chcesz zalogować się jeszcze raz?
Szalki petriego, Gosselin™
Dostawca: Corning Life Sciences
Gosselin
Transparent PS Petri dishes for food industry laboratories.
  • Excellent optical quality
  • Stable stacking due to the stacking ring
  • High level of mechanical resistance, will retain shape up to +55 °C
These dishes are produced in a Class 100 (ISO 5) manufacturing environment, ensuring an aseptic product. Product is labelled with a batch number on each carton. Also available sterile (Sterility Assurance Level based on ISO 11137 : SAL 10-6).
ZAMÓW
Rozwiń / Zwiń
Aseptic
Wersja Ø Wysokość Ilość Numer katalogowy VWR Ilość
With triple vents 55 mm 14,2 mm 1.620 391-0570
With triple vents 90 mm 14,2 mm 825 391-0572
2 compartments, 3 vents 90 mm 14,2 mm 825 391-0567
Without vents 90 mm 14,2 mm 825 391-0571
With triple vents 100 mm 14,2 mm 825 GOSSBP93B-101
Sterile
Wersja Ø Wysokość Sterylność Ilość Numer katalogowy VWR Ilość
With triple vents 55 mm 14,2 mm + 1.620 391-0569
With triple vents 90 mm 14,2 mm + 825 391-0568
With triple vents 90 mm 16,2 mm + 700 391-0566
Without vents 90 mm 14,2 mm + 825 GOSSBP90B-102
With triple vents 100 mm 14,2 mm + 825 GOSSBP93D-102
With triple vents 100 mm 14,2 mm + 825 GOSSSB93-102
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość