Drums with lid and clamping ring
: Kautex
HDPE. With lid and clamping ring.
- Lid
- Clamping ring
- Suitable for food
- All sizes with UN approval for solids
- Halal certified
Barrels meet the GGVS/GGVE (rail/road/sea) regulations for the transport of solid substances with a density of at least 1,5 g/ccm. Supplied complete with PE lid and galvanized clamping ring. 30 and 60 L with handles.
- CapacityExt.ØØ openingHeightPk
- UN approval: UN 1H2/X61/SUN approval: UN 1H2/X61/S216-5652359-9080030 L313 mm251 mm518 mm11 * 1 SZT365 zł
Specifications
Environmentally Preferable - UN approval: UN 1H2/X88/SUN approval: UN 1H2/X88/S216-5541359-9086760 L398 mm325 mm620 mm11 * 1 SZT506 zł
Specifications
Environmentally Preferable - UN approval: UN 1H2/X225/SUN approval: UN 1H2/X225/S216-5542359-91093120 L469 mm395 mm790 mm11 * 1 SZT749 zł
Specifications
Environmentally Preferable