Drukuj
Wyszukano: Zatyczki do uszu
62  wyniki/ów wyszukiwania
SearchResultCount:"62"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: 3M
Opis: The unique, one size E-A-R™ Tracer™ earplugs fit most ear canals comfortably and provide high levels of attenuation with the added benefit of incorpor...
Producent: 3M
Opis: This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industr...
Producent: 3M
Opis: These earplugs are made from flexible materials that are already pre-formed to ensure optimum fit.
Producent: 3M
Opis: E-A-R™ Flexible Fit HA earplugs have a patented earplug foam formulation which makes them fully washable and reusable. The soft foam tip and flexible ...
Numer katalogowy: (MMMAUF01012)
Producent: 3M
Opis: These pre-moulded earplugs are suitable for moderate noise levels (83 to 93 dB(A)). Correctly matched to the noise level they allow conversation and b...
j.m.: 1 * 50 PARA
Numer katalogowy: (MMMAUF01015)
Producent: 3M
Opis: These pre-moulded earplugs made from soft TPE use unique filter technology to take the edge off low levels of harmful noise whilst allowing normal con...
j.m.: 1 * 200 PARA
Producent: 3M
Opis: E-A-RSoft™ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexib...
Numer katalogowy: (MMMAUF01000S)
Producent: 3M
Opis: The 3M™ E-A-R™ UltraFit™ Earplugs are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up t...
j.m.: 1 * 200 PARA
Producent: 3M
Opis: Earplugs manufactured from a soft elastic material to ensure a secure and comfortable fit.
Producent: 3M
Opis: E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can re...
Producent: 3M
Opis: Te rolowane zatyczki do uszu produkowane są z miękkiej, pochłaniającej energię pianki polimerowej, która zapewnia doskonałą ochronę słuchu i wygodę pr...
Producent: 3M
Opis: Soft, smooth, dirt resistant foam earplugs mould to the unique shape of each ear for excellent hearing protection.
Producent: 3M
Opis: This lightweight semi-aural hearing protector is ideal when moving in and out of high noise areas, fitting comfortably around the neck when not in use...
Producent: 3M
Opis: These pod earplugs are very easy to insert since there is no roll needed, users can just use the soft flexible grip to push the earplug into position....
Producent: 3M
Opis: These earplugs are ideal for long periods of use with changing noise levels.
Numer katalogowy: (MMMAEC01000S)
Producent: 3M
Opis: The E-A-Rcaps™ banded hearing protector is fitted with semi-aural eartips designed to seal the entrance of the ear canal to help reduce exposure to ha...
j.m.: 1 * 400 PARA
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 62
no targeter for Bottom