Centrifugal filtration units, Vivaspin® 500
Supplier: Sartorius
Certificates
About this item
Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filter units offer a simple, one step procedure for sample concentration with volumes from 5 to 500 µl. They can effectively be used in fixed angle rotors accepting 2,2 ml centrifuge tubes.
- High retentate recovery >90%
- Vertical membrane design for ultra-fast concentration
- Safe sample concentration due to dead-stop volume
- Convenient concentrate recovery by pipetting
- Volume graduation for easy reading/monitoring
The patented vertical membrane design and thin channel filtration chamber (US 5,647,990), minimises membrane fouling and provides high speed concentrations.
Specifications
- Catalog No:
- 512-3737
- 512-4026
- 512-4005
- 512-3736
- 512-4025
- 512-4004
- 512-4003
- 512-3744
- 512-3739
- 512-3738
- 512-3740
- 512-2838
- 512-2837
- 512-4002
- 512-4001
- Filtration area:
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- Max. RCF:
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- 15000 ×g
- Dead volume:
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- Ø×H:
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
Specifications
Product Information
- Catalog #Supplier #MWCOPore sizeMembranePkAvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Environmentally PreferableFiltration area0,5 cm²Max. RCF15000 ×gDead volume<5 µlØ×H11×50 mm