Volumetric flasks, class A, blue markings
About this item
Related Information
Certificates
Search by Lot NumberHirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
With batch certificate
Product Information
- Catalog #Supplier #CapacityNSTolerancePkAvailabilityPrice
- 612-155628201525 ml7/160,025 ± ml21 * 2 SZT174 złSupplier Article Number: 2820152Local Article Number: HIRS2820152Specifications:
Specifications
Environmentally Preferable - 612-1558282016010 ml7/160,025 ± ml21 * 2 SZT174 złSupplier Article Number: 2820160Local Article Number: HIRS2820160Specifications:
Specifications
Environmentally Preferable - 612-1562282016520 ml10/190,040 ± ml21 * 2 SZT181 złSupplier Article Number: 2820165Local Article Number: HIRS2820165Specifications:
Specifications
Environmentally Preferable - 612-1563282017025 ml10/190,040 ± ml21 * 2 SZT181 złSupplier Article Number: 2820170Local Article Number: HIRS2820170Specifications:
Specifications
Environmentally Preferable - 612-1564282017125** ml12/210,060 ± ml21 * 2 SZT200 złSupplier Article Number: 2820171Local Article Number: HIRS2820171Specifications:
Specifications
Environmentally Preferable - 612-1565282017550 ml12/210,060 ± ml21 * 2 SZT198 złSupplier Article Number: 2820175Local Article Number: HIRS2820175Specifications:
Specifications
Environmentally Preferable - 612-1566282017850** ml14/230,080 ± ml21 * 2 SZT201 złSupplier Article Number: 2820178Local Article Number: HIRS2820178Specifications:
Specifications
Environmentally Preferable - 612-15672820180100 ml12/210,100 ± ml21 * 2 SZT219 złSupplier Article Number: 2820180Local Article Number: HIRS2820180Specifications:
Specifications
Environmentally Preferable - 612-15682820181100 ml14/230,100 ± ml21 * 2 SZT221 złSupplier Article Number: 2820181Local Article Number: HIRS2820181Specifications:
Specifications
Environmentally Preferable - 612-15692820185200 ml14/230,150 ± ml21 * 2 SZT248 złSupplier Article Number: 2820185Local Article Number: HIRS2820185Specifications:
Specifications
Environmentally Preferable - 612-15702820186250 ml14/230,150 ± ml21 * 2 SZT256 złSupplier Article Number: 2820186Local Article Number: HIRS2820186Specifications:
Specifications
Environmentally Preferable - 612-15712820190500 ml19/260,250 ± ml21 * 2 SZT305 złSupplier Article Number: 2820190Local Article Number: HIRS2820190Specifications:
Specifications
Environmentally Preferable - 612-157228201921000** ml29/320,600 ± ml11 * 1 SZT286 złSupplier Article Number: 2820192Local Article Number: HIRS2820192Specifications:
Specifications
Environmentally Preferable - 612-157428201942000 ml29/320,600 ± ml11 * 1 SZT360 złSupplier Article Number: 2820194Local Article Number: HIRS2820194Specifications:
Specifications
Environmentally Preferable - 612-330728201975000 ml34/351,200 ± ml11 * 1 SZT757 złSupplier Article Number: 2820197Local Article Number: HIRS2820197Specifications:
Specifications
Environmentally Preferable - 612-155728201535** ml10/190,040 ± ml21 * 2 SZT174 złSupplier Article Number: 2820153Local Article Number: HIRS2820153Specifications:
Specifications
Environmentally Preferable - 612-1559282016110** ml10/190,040 ± ml21 * 2 SZT174 złSupplier Article Number: 2820161Local Article Number: HIRS2820161Specifications:
Specifications
Environmentally Preferable - 612-5737282016620** ml12/210,040 ± ml11 * 1 SZT114 złSupplier Article Number: 2820166Local Article Number: HIRS2820166Specifications:
Specifications
Environmentally Preferable - 612-58722820182100 ml19/260,100 ± ml21 * 2 SZT343 złSupplier Article Number: 2820182Local Article Number: HIRS2820182Specifications:
Specifications
Environmentally Preferable - 612-157328201931000 ml24/290,400 ± ml11 * 1 SZT264 złSupplier Article Number: 2820193Local Article Number: HIRS2820193Specifications:
Specifications
Environmentally Preferable - 612-3308282019810000* ml45/402,000 ± ml11 * 1 SZT1 460 złSupplier Article Number: 2820198Local Article Number: HIRS2820198Specifications:
Specifications
Environmentally Preferable
With individual certificate
Product Information
- Catalog #Supplier #CapacityNSTolerancePkAvailabilityPrice
- 612-586928201780850** ml14/230,080 ± ml11 * 1 SZT233 złSupplier Article Number: 282017808Local Article Number: HIRS282017808Specifications:
Specifications
Environmentally Preferable - 612-58632820152085 ml7/160,025 ± ml11 * 1 SZT219 złSupplier Article Number: 282015208Local Article Number: HIRS282015208Specifications:
Specifications
Environmentally Preferable - 612-586428201600810 ml7/160,025 ± ml11 * 1 SZT219 złSupplier Article Number: 282016008Local Article Number: HIRS282016008Specifications:
Specifications
Environmentally Preferable - 612-586528201610810** ml10/190,040 ± ml11 * 1 SZT219 złSupplier Article Number: 282016108Local Article Number: HIRS282016108Specifications:
Specifications
Environmentally Preferable - 612-586628201650820 ml10/190,040 ± ml11 * 1 SZT223 złSupplier Article Number: 282016508Local Article Number: HIRS282016508Specifications:
Specifications
Environmentally Preferable - 612-586728201700825 ml10/190,040 ± ml11 * 1 SZT223 złSupplier Article Number: 282017008Local Article Number: HIRS282017008Specifications:
Specifications
Environmentally Preferable - 612-586828201750850 ml12/210,060 ± ml11 * 1 SZT231 złSupplier Article Number: 282017508Local Article Number: HIRS282017508Specifications:
Specifications
Environmentally Preferable - 612-5870282018008100 ml12/210,100 ± ml11 * 1 SZT241 złSupplier Article Number: 282018008Local Article Number: HIRS282018008Specifications:
Specifications
Environmentally Preferable - 612-5871282018108100 ml14/230,100 ± ml11 * 1 SZT243 złSupplier Article Number: 282018108Local Article Number: HIRS282018108Specifications:
Specifications
Environmentally Preferable - 612-5873282018508200 ml14/230,150 ± ml11 * 1 SZT256 złSupplier Article Number: 282018508Local Article Number: HIRS282018508Specifications:
Specifications
Environmentally Preferable - 612-58782820194452000 ml29/320,600 ± ml11 * 1 SZT535 złSupplier Article Number: 282019445Local Article Number: HIRS282019445Specifications:
Specifications
Environmentally Preferable - 612-58792820197085000 ml34/351,200 ± ml11 * 1 SZT890 złSupplier Article Number: 282019708Local Article Number: HIRS282019708Specifications:
Specifications
Environmentally Preferable - 612-588028201980810000* ml45/402,000 ± ml11 * 1 SZT1 590 złSupplier Article Number: 282019808Local Article Number: HIRS282019808Specifications:
Specifications
Environmentally Preferable - 612-5874282018608250 ml14/230,150 ± ml11 * 1 SZT260 złSupplier Article Number: 282018608Local Article Number: HIRS282018608Specifications:
Specifications
Environmentally Preferable - 612-5875282019008500 ml19/260,250 ± ml11 * 1 SZT284 złSupplier Article Number: 282019008Local Article Number: HIRS282019008Specifications:
Specifications
Environmentally Preferable - 612-58762820193081000 ml24/290,400 ± ml11 * 1 SZT396 złSupplier Article Number: 282019308Local Article Number: HIRS282019308Specifications:
Specifications
Environmentally Preferable - 612-58772820194082000 ml29/320,600 ± ml11 * 1 SZT492 złSupplier Article Number: 282019408Local Article Number: HIRS282019408Specifications:
Specifications
Environmentally Preferable