Safety waste systems, Nalgene®
About this item
HDPE.
- 10 L made from fluorinated HDPE, 4 L from HDPE
- PP funnel includes cover with easy snap safety latch to minimise accidental spills and volatile emissions
- Built in vent plug releases pressure during pouring and prevents emissions during use
- Closure/adapter can also be connected to an external syringe filter
- Removable PP screen insert/filter fits at bottom of funnel to trap large particles
A system for temporary storage of solvent, chemical, and biological waste which consists of a chemical and break resistant container and removable funnel with lid, closure/funnel adapter, and screen insert.
- Catalog No:
- 119-3010
- 119-3004
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Safety waste system (container and funnel)
- CapacityNeck-ØCap SizePk
- HDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insertHDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insert119-30106379-001010 L250 mm83B11 * 1 SZT1 500 zł
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - HDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insertHDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insert119-30046379-00044 L140 mm38-43011 * 1 SZT964 zł
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact
Safety funnels
- ThreadCapacityFunnel O-ØStem lengthPkPrice