Drukuj
System turbodołądowania powietrza, Jupiter™
Dostawca: 3M
Jupiter™
111-0240EA 5720 PLN
111-0240
System turbodołądowania powietrza, Jupiter™
Maski oddechowe Systemy Oddechowe
Low profile, modular, powered air purifying respirator for use with quick release (QRS) helmets. Wide choice of filters for gases, vapours and particulates, with a filter cover (easy to clean) to reduce dust and prevent spark damage.
  • Comfortable and flexible: Freedom of movement especially in smaller spaces
  • Easy to use and maintain
  • The ergonomic comfort belt remains comfortable even when wearing for a longer period of time
  • The electronic alarm gives a warning (audible and visual) of low airflow and/or low battery
The standard Jupiter™ Powered Air Base Unit includes decontamination belt, airflow indicator and calibration tube.
Informacje do zamówienia: Versaflo™ S-Series helmets sold separately.