Robotic tips, Axygen®, for PerkinElmer® Janus®, Evolution P3, MiniTrak & PlateTrak®
Supplier: Corning
Certificates
About this item
Free of RNase, DNase, DNA and pyrogens.
- Transparent PP
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any pipette contamination
- Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
Specifications
- Catalog No:
- 736-0465
- 736-0469
- 736-0294
- 613-0968
- 736-0299
- 736-0296
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Information
- Catalog #Supplier #Tip styleVolumeSterileFilterTip colourFor use withAvailabilityPrice
- 736-0294PK-20-RPK-20-R20 µl--TransparentMDT-96 P501 * 960 SZT472 złSpecifications:
Specifications
Environmentally PreferableEnvironmental certification - 613-0968PK-384-RPK-384-R30 µl--TransparentMDT-384-30 µl1 * 3.840 SZT4 490 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 736-0296PK-50-RPK-50-R50 µl--TransparentMDT-96 P501 * 960 SZT466 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 736-0299PK-200-RPK-200-R200 µl--TransparentMDT-96 P2351 * 960 SZT543 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 736-0465PK-384-R-SPK-384-R-S30 µl+-TransparentMDT-384-30 µl1 * 3.840 SZT4 590 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 736-0469PKF-384-R-SPKF-384-R-S30 µl++TransparentMDT-384-30 µl1 * 3.840 SZT5 190 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact