Drukuj
Okulary ochronne, Cybric
Dostawca: Uvex
uvex cybric
111-0146EA 170 PLN
111-0146 111-1655 111-1657
Okulary ochronne, Cybric
Okulary
Sporty designed dual lens safety glasses with innovative lenses.
  • Padded arms ensure a pressure-free and comfortable fit
  • Wide field of vision
Weight: 48 g
Certyfikaty: EN 166, EN 168, EN 170