Drukuj
Absorbent do rurek dializowania Spectra/Gel®
Dostawca: Spectrum
734-0858EA 763 PLN
SPEC292600
Absorbent do rurek dializowania Spectra/Gel®
Spectra/Gel® makes the concentration of protein sample solutions quick and efficient. Made of polyacrylate-polyalcohol it has the property of expanding as it absorbs water. The high molecular weight polymer can be used with dialysis membranes of molecular weight cut-offs (MWCO) from 100 to 300000 Daltons without contaminating the sample. After dialysis, simply remove the dialysis tubing containing the sample and coat it with Spectra/Gel®. Water and smaller molecules diffuse through the membrane and are absorbed by Spectra/Gel® Absorbent. Concentration continues unattended until a certain volume of water is removed from the sample. The insoluble characteristic makes Spectra/Gel® easy to remove after absorbancy.
  • High water absorbent capacity
  • Easy to use with any MWCO dialysis membrane
  • Easy to concentrate small molecular weight molecules
  • Convenient for sample concentration with Float-A-Lyzer® G2