Order Entry
VWR® ND13, Screw Cap PP with Central Hole, 8,5 mm and Septa
VWR® ND13, Screw Cap PP with Central Hole, 8,5 mm and Septa
undefined
VWR® ND13, Screw Cap PP with Central Hole, 8,5 mm and Septa
  •  
    Type
    Hardness
    Septum
    Thickness
    Pk
  • 548-3054
    13150815
    PP black
    55° shore A
    Silicone beige/PTFE red
    1,5 mm
    100
    1 * 100 SZT
    1 * 100 SZT - 391 zł
    391 zł
    1 * 100 SZT
       391 zł
    Specifications
    Environmentally Preferable
  • 548-3095
    13150439
    PP black, closed top
    60° shore A
    Natural rubber red-orange/TEF transparent
    1,3 mm
    100
    1 * 100 SZT
    1 * 100 SZT - 249 zł
    249 zł
    1 * 100 SZT
       249 zł
    Specifications
    Environmentally Preferable
  • 548-3053
    13150456
    PP black
    60° shore A
    Natural rubber red-orange/TEF transparent
    1,3 mm
    100
    1 * 100 SZT
    1 * 100 SZT - 242 zł
    242 zł
    1 * 100 SZT
       242 zł
    Specifications
    Environmentally Preferable