Ubbelohde viscometers, DIN
Supplier: SI Analytics
Certificates
About this item
Ubbelohde viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
- Additional filling and cleaning tube
- Glass threads ensure safe operational use
- Calibrated with constant for automatic measurement
Certifications: DIN 51562-1, ISO 3105, BS 133, NFT 60-100
Specifications
- Catalog No:
- 616-3380
- 616-3385
- 616-3384
- 616-3387
- 616-3383
- 616-3382
- Length:
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
Specifications
Product Information
- Catalog #Supplier #RangeConstantLengthØ int.VolumePkAvailabilityPrice
- 616-33802854019250,5 - 3 mm²/s0,003 K290 mm0,47 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable - 616-33822854019413 - 30 mm²/s0,03 K290 mm0,84 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable - 616-338328540195810 - 100 mm²/s0,1 K290 mm1,13 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable - 616-338428540196630 - 300 mm²/s0,3 K290 mm1,50 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable - 616-3385285401974100 - 1000 mm²/s1 K290 mm2,01 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable - 616-33872854019991000 - 6000 mm²/s10 K290 mm3,70 mm18 − 22 ml1Log in for availability1 * 1 SZT3 370 złSpecifications:
More
Specifications
Length290 mmEnvironmentally Preferable