ScrubPADS UltraSOLV® Chamber Cleaning Pads
: FOAMTEC
Silicon carbide ScrubPADS
- L×WPk
- 600-grit: HT4560-10: FOAMHT4560-10
Specifications
Environmentally Preferable - 280-Grit: HT4528-11: FOAMHT4528-11
Specifications
Environmentally Preferable - 1500-grit: HT4515P-10: FOAMHT4515P-10
Specifications
Environmentally Preferable - 400-grit: HT4540-10: FOAMHT4540-10
Specifications
Environmentally Preferable - 220-grit: HT4522-10: FOAMHT4522-10
Specifications
Environmentally Preferable - 100-grit: HT4510-10: FOAMHT4510-10
Specifications
Environmentally Preferable - 280-grit: HT4528-10: FOAMHT4528-10
Specifications
Environmentally Preferable - 800-grit: HT4580-10: FOAMHT4580-10
Specifications
Environmentally Preferable - 800-grit: HT4580-11: FOAMHT4580-11
Specifications
Environmentally Preferable - 1200-grit: HT4512P-10: FOAMHT4512P-10
Specifications
Environmentally Preferable
Aluminium oxide ScrubPADS
- L×WPk
- 220-grit: HT4522A-10: FOAMHT4522A-10
Specifications
Environmentally Preferable - 1200-grit1200-grit115-0398HT4512PA-10102×76 mm101 * 10 SZT15,80 zł: HT4512PA-10: FOAMHT4512PA-10
Specifications
Environmentally Preferable - 800-grit: HT4580A-10: FOAMHT4580A-10
Specifications
Environmentally Preferable
Diamond ScrubPADS
- L×WPk
- 1350-grit: HT4513PD-10: FOAMHT4513PD-10
Specifications
Environmentally Preferable - 280-Grit: HT4528D-10: FOAMHT4528D-10
Specifications
Environmentally Preferable - 360-grit: HT4536D-10: FOAMHT4536D-10
Specifications
Environmentally Preferable - 400-grit: HT4540D-10: FOAMHT4540D-10
Specifications
Environmentally Preferable
Foam wiping products
- L×WPk
- Rounded tip: HT1702-50: FOAMHT1702-50
Specifications
Environmentally Preferable - Medium flex-tip: HT1701-5: FOAMHT1701-5
Specifications
Environmentally Preferable - Rigid rounded tip, UltraSOLV® foam headRigid rounded tip, UltraSOLV® foam head115-0392HT1732-5011×3,6 mm501 * 50 SZT2 zł: HT1732-50: FOAMHT1732-50
Specifications
Environmentally Preferable - Rigid spear tip: HT1714-50: FOAMHT1714-50
Specifications
Environmentally Preferable - Large bulb tip: HT1700-5: FOAMHT1700-5
Specifications
Environmentally Preferable