Drukuj
Wyszukano: Analizatory
27  wyniki/ów wyszukiwania
SearchResultCount:"27"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Collection
Opis: Moisture analysers with large colour touch screen display give a user-friendly access to advanced applications and internal databases. The user has an...
Producent: VWR Collection
Opis: Rugged compact moisture analysers with large backlit dot matrix graphic display for moisture determination of raw materials and finished products in t...
Numer katalogowy: (620-1956)
Producent: VWR Collection
Opis: The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or...
j.m.: 1 * 1 SZT
Producent: Sartorius Balances
Opis: The MA100 is an accurate and configurable model with a user friendly keyboard and software prompts to easily access all the modes of operation and dat...
Numer katalogowy: (611-4357)
Producent: Sartorius Balances
Opis: This infrared moisture analyser is designed for use in daily routine in-process control.
j.m.: 1 * 1 SZT
Numer katalogowy: (611-2887)
Producent: Sartorius Balances
Opis: Łatwe w użyciu, wydajne, kompaktowe analizatory wilgotności zapewniające szybkie i powtarzalne wyniki z ulepszonym interfejsem użytkownika.
j.m.: 1 * 1 SZT
Numer katalogowy: (611-2888)
Producent: Sartorius Balances
Opis: Compact moisture analyser giving fast repeatable results with an improved user interface.
j.m.: 1 * 1 SZT
Producent: OHAUS
Opis: Providing accurate and reproducible results at either 0,1% or 0,05% readability.
Producent: OHAUS
Opis: Moisture analysers simplify complex moisture analysis applications. Designed to increase precision and efficiency in the laboratory, they feature a pr...
Numer katalogowy: (SIMELI0015)
Producent: Industrial Physics
Opis: Combiscan is a pinless moisture meter that offers analysis of 30 different wood species and more than 240 wood types by 15 density groups (0,3 to 1,0)...
j.m.: 1 * 1 SZT
Producent: Industrial Physics
Opis: profi LIGNO are moisture meters specifically designed for wood and building materials such as screed or gypsum.
Producent: Industrial Physics
Opis: These hardness tests are pocket instruments for testing the hardness and wear/scratch resistance of materials such as coatings, lacquers, plastics or ...
Numer katalogowy: (SIMELI9200)
Producent: Industrial Physics
Opis: This meter is a non-destructive moisture meter for concrete. By measuring the electrical impedance, the moisture content of concrete can be determined...
j.m.: 1 * 1 SZT
Numer katalogowy: (SIMELI0006)
Producent: Industrial Physics
Opis: The mini LIGNO classic is a wood moisture meter that offers the choice between four different base-wood groups.
j.m.: 1 * 1 SZT
Numer katalogowy: (SIMELI7000)
Producent: Industrial Physics
Opis: Non destructive instrument for measuring moisture levels in wood.
j.m.: 1 * 1 SZT
Numer katalogowy: (SIMELI0004)
Producent: Industrial Physics
Opis: mini LIGNO duo is a wood moisture meter that provides four different base-wood groups, eight building materials, such as gypsum or screed.
j.m.: 1 * 1 SZT
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 27
no targeter for Bottom