Drukuj
Wyszukano: Bloki grzejne, cyfrowe
26  wyniki/ów wyszukiwania
SearchResultCount:"26"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Collection
Opis: Wielozadaniowe cyfrowe grzejniki blokowe VWR® o wysokiej mocy i stałej temperaturze są ekonomiczne, wszechstronne i kompaktowe. Te cyfrowe modele zape...
Zmniejszanie ilości odpadów
Producent: VWR Collection
Opis: These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space....
Zmniejszanie ilości odpadów
Numer katalogowy: (460-0353)
Producent: VWR Collection
Opis: Wielofunkcyjny, o wysokiej mocy i stałej temperaturze cyfrowy grzejnik blokowy VWR®, który jest ekonomiczny, wszechstronny i kompaktowy. Ten cyfrowy m...
j.m.: 1 * 1 SZT
Zmniejszanie ilości odpadów
Producent: GRANT INSTRUMENTS LTD.
Opis: Compact digital block heaters with choice of models for 1 or 2 or 4 interchangeable blocks. Units feature an LED temperature display, easy to use inte...
Numer katalogowy: (460-0267)
Producent: GRANT INSTRUMENTS LTD.
Opis: Compact digital block heater with fixed circular block. Unit features digital control, two line display allowing simple and accurate setting of temper...
j.m.: 1 * 1 SZT
Producent: GRANT INSTRUMENTS LTD.
Opis: High temperature digital block heaters with single fixed aluminium block. Units feature microprocessor control, digital temperature settings and displ...
Producent: 2MAG
Opis: Ideal for viscosity measurements, testing and quality control. The units have a clear digital display for settings of stirrer speed, stirrer power and...
Numer katalogowy: (TMAG63825)
Producent: 2MAG
Opis: The aluminium alloy block heater is PTFE coated for chemical resistance and easy cleaning. The combined stirring block heater has 8 stirring positions...
j.m.: 1 * 1 SZT
Producent: WTW
Opis: Thermoreactors for the determination of COD, total nitrogen or total phosphorus, ensure complete digestion of the sample and maintain the necessary hi...
Producent: IKA
Opis: Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sen...
Numer katalogowy: (LOVI2418940-V)
Producent: Lovibond
Opis: The RD 125 thermoreactor can be used for the tube test digestion of highly sensitive and precise COD (150 °C), TOC (120 °C), total chromium (100 °C), ...
j.m.: 1 * 1 SZT
Numer katalogowy: (614-0200)
Producent: GERBER FUNKE DR N.
Opis: The RD-8 was specifically developed in conjunction with COKZ (the Netherlands Controlling Authority for Milk and Milk products) for determining the mo...
j.m.: 1 * 1 SZT
Producent: BARNSTEAD (INC THERMOLYNE)
Opis: Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducibl...
Producent: BARNSTEAD (INC THERMOLYNE)
Opis: Designed to provide precise temperature stability and uniformity. The units have an easy to use touch screen control which displays time and temperatu...
Producent: Cole Parmer
Opis: Provides precise heating of tubes and microplates.
Producent: OHAUS
Opis: Designed for applications that require repeatable results and superior temperature stability. These multi-purpose units are ideal for incubation and a...
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 26
no targeter for Bottom