Drukuj
Kolorymetr, Spectroquant® Multy
Dostawca: Merck
1.73630.0001EA 6280 PLN
MERC1.73630.0001
Kolorymetr, Spectroquant® Multy
The Spectroquant® Multy is the ideal instrument for an inexpensive yet wide ranging introduction to photometric water analysis. The equipment is pre-programmed for the use of over 130 Spectroquant® cell and reagent tests and covers all the essential parameters of wastewater and drinking water analysis, e.g. COD, total nitrogen, total phosphorus and many others. Also trace levels of cyanide, arsenic, cadmium and other materials can be measured very precisely. The broad range of easy to handle Spectroquant® test kits offers an optimal solution for each user. Rechargeable batteries are included, and together with the practical carrying case, enable the instrument to be used anywhere, anytime. Methods can be updated free of charge over the Internet.
  • Easy to use
  • Water resistant instrument (whilst not charging)
  • Robust construction for field or laboratory use
  • Battery recharging whilst using the instrument via integrated charger
For full range of tests see table ‘Test Kits, Spectroquant®'.
Dodatkowe informacje: Multy colorimeter, case, adapter unit for 16 mm round cells, lid for adapter unit, seven rechargeable batteries, lithium battery (to ensure data storage) interface cable for connection to PC, 3×16 mm round cells, 3×24 mm round cells.