Dish-shaped Dewar flasks, type SCH-CAL
Supplier: KGW
Certificates
About this item
Silvered borosilicate glass, structured aluminium protective covering. For tempering of round bottom flasks, cooling of small samples. They are vacuum insulated and silvered.
- Suitable as hot or cold bath
- Can be used on a magnetic stirrer
- The cover is anti-magnetic and stainless
Temperature range from −200 to +200 °C.
CAL flasks have an external stucco aluminium protective casing, alternatively the glass inserts (without casing) are also available.
Specifications
- Catalog No:
- 478-4511
- 478-4507
- 478-4510
- 478-4506
- KGWA1671
- KGWA1675
- KGWA1674
- 478-4505
- 478-4503
- 478-0226
- KGWA1673
- 478-4509
- KGWA1672
- Model:
- SCH 33 CAL
- SCH 20 CAL
- SCH 31 CAL
- SCH 18 CAL
- SCH 15 CAL
- SCH 9 CAL
- SCH 6 CAL
- SCH 30 CAL
- Capacity:
- 5,3 L
- 0,80 L
- 2,7 L
- 0,40 L
- 0,26 L
- 0,12 L
- 0,08 L
- 1,6 L
- Temperature range:
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- −200...+200 °C
- Int.Ø:
- 250 mm
- 138 mm
- 200 mm
- 110 mm
- 100 mm
- 77 mm
- 67 mm
- 170 mm
- Int. height:
- 150 mm
- 80 mm
- 125 mm
- 70 mm
- 65 mm
- 50 mm
- 40 mm
- 110 mm
- Ext.Ø:
- 280 mm
- 160 mm
- 230 mm
- 130 mm
- 120 mm
- 95 mm
- 80 mm
- 205 mm
- Overall height:
- 195 mm
- 125 mm
- 160 mm
- 115 mm
- 110 mm
- 90 mm
- 75 mm
- 145 mm
Specifications
Product Information
- Catalog #Supplier #ForPkAvailabilityPrice
- Dish-shaped Dewar flask, SCH 6 CALDish-shaped Dewar flask, SCH 6 CAL478-0226119180 ml round bottom flasks (67 mm Ø)1Log in for availability1 * 1 SZT951 złSupplier Article Number: 1191Local Article Number: KGWA1191Specifications:
More
Accessories
Specifications
ModelSCH 6 CALCapacity0,08 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø67 mmInt. height40 mmExt.Ø80 mmOverall height75 mm - Dish-shaped Dewar flask, SCH 9 CALDish-shaped Dewar flask, SCH 9 CAL478-45031192120 ml round bottom flasks (77 mm Ø)1Log in for availability1 * 1 SZT1 020 złSupplier Article Number: 1192Local Article Number: KGWA1192Specifications:
More
Accessories
Specifications
ModelSCH 9 CALCapacity0,12 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø77 mmInt. height50 mmExt.Ø95 mmOverall height90 mm - Dish-shaped Dewar flask, SCH 15 CALDish-shaped Dewar flask, SCH 15 CAL478-45051193260 ml round bottom flasks (100 mm Ø)1Log in for availability1 * 1 SZT1 430 złSupplier Article Number: 1193Local Article Number: KGWA1193Specifications:
More
Accessories
Specifications
ModelSCH 15 CALCapacity0,26 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø100 mmInt. height65 mmExt.Ø120 mmOverall height110 mm - Dish-shaped Dewar flask, SCH 18 CALDish-shaped Dewar flask, SCH 18 CAL478-45061194400 ml round bottom flasks (110 mm Ø)1Log in for availability1 * 1 SZT1 710 złSupplier Article Number: 1194Local Article Number: KGWA1194Specifications:
More
Accessories
Specifications
ModelSCH 18 CALCapacity0,40 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø110 mmInt. height70 mmExt.Ø130 mmOverall height115 mm - Dish-shaped Dewar flask, SCH 20 CALDish-shaped Dewar flask, SCH 20 CAL478-45071195800 ml round bottom flasks (138 mm Ø)1Log in for availability1 * 1 SZT2 010 złSupplier Article Number: 1195Local Article Number: KGWA1195Specifications:
More
Accessories
Specifications
ModelSCH 20 CALCapacity0,80 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø138 mmInt. height80 mmExt.Ø160 mmOverall height125 mm - Dish-shaped Dewar flask, SCH 30 CALDish-shaped Dewar flask, SCH 30 CAL478-450911961600 ml round bottom flasks (170 mm Ø)1Log in for availability1 * 1 SZT2 840 złSupplier Article Number: 1196Local Article Number: KGWA1196Specifications:
More
Accessories
Specifications
ModelSCH 30 CALCapacity1,6 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø170 mmInt. height110 mmExt.Ø205 mmOverall height145 mm - Dish-shaped Dewar flask, SCH 31 CALDish-shaped Dewar flask, SCH 31 CAL478-451011972700 ml round bottom flasks (200 mm Ø)1Log in for availability1 * 1 SZT4 280 złSupplier Article Number: 1197Local Article Number: KGWA1197Specifications:
More
Accessories
Specifications
ModelSCH 31 CALCapacity2,7 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø200 mmInt. height125 mmExt.Ø230 mmOverall height160 mm - Dish-shaped Dewar flask, SCH 33 CALDish-shaped Dewar flask, SCH 33 CAL478-451111985300 ml round bottom flasks (250 mm Ø)1Log in for availability1 * 1 SZT7 390 złSupplier Article Number: 1198Local Article Number: KGWA1198Specifications:
More
Accessories
Specifications
ModelSCH 33 CALCapacity5,3 LEnvironmentally PreferableTemperature range−200...+200 °CInt.Ø250 mmInt. height150 mmExt.Ø280 mmOverall height195 mm
Lid
Product Information
- Catalog #Supplier #ForPkAvailabilityPrice
- Dewar lid for SCH 9 CAL vesselDewar lid for SCH 9 CAL vesselKGWA16711671Dewar flask SCH 9 CAL1Log in for availability1 * 1 SZT293 złSupplier Article Number: 1671Local Article Number: KGWA1671Specifications:
Accessories
Specifications
ModelCapacityEnvironmentally PreferableTemperature rangeInt.ØInt. heightExt.ØOverall height - Dewar lid for SCH 15 CAL vesselDewar lid for SCH 15 CAL vesselKGWA16721672Dewar flask SCH 15 CAL1Log in for availability1 * 1 SZT310 złSupplier Article Number: 1672Local Article Number: KGWA1672Specifications:
Accessories
Specifications
ModelCapacityEnvironmentally PreferableTemperature rangeInt.ØInt. heightExt.ØOverall height - Supplier Article Number: 1673Local Article Number: KGWA1673Specifications:
Accessories
Specifications
ModelCapacityEnvironmentally PreferableTemperature rangeInt.ØInt. heightExt.ØOverall height - Dewar lid for SCH 20 CAL vesselDewar lid for SCH 20 CAL vesselKGWA16741674Dewar flask SCH 20 CAL1Log in for availability1 * 1 SZT364 złSupplier Article Number: 1674Local Article Number: KGWA1674Specifications:
Accessories
Specifications
ModelCapacityEnvironmentally PreferableTemperature rangeInt.ØInt. heightExt.ØOverall height - Dewar lid for SCH 30 CAL vesselDewar lid for SCH 30 CAL vesselKGWA16751675Dewar flask SCH 30 CAL1Log in for availability1 * 1 SZT815 złSupplier Article Number: 1675Local Article Number: KGWA1675Specifications:
Accessories
Specifications
ModelCapacityEnvironmentally PreferableTemperature rangeInt.ØInt. heightExt.ØOverall height
Glass refills
Product Information
- Catalog #SeqDescriptionPkAvailabilityPrice
- Supplier Article Number: 1175Local Article Number: KGWA1175