Order Entry
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Supplier:  WTW ONLINE
Certificates
undefined
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Supplier:  WTW ONLINE
Specifications
  • Catalog No:
  • 706-0524
  • 706-0525
  • Model:
  • FDO 700 IQ
  • FDO 700 IQ SW
  • Min. flow rate:
  • No flow required
  • No flow required
  • Response time:
  • <150 s
  • <60 s
  • DO concentration:
  • 0...20,00 mg/l
  • 0...20,00 mg/l
  • DO saturation:
  • 0...200,0%
  • 0...200,0%
  • DO barometric pressure range:
  • 10 bar
  • 10 bar
  • DO accuracy:
  • ±0,05 mg/l
  • ±0,05 mg/l
  • DO resolution:
  • 0,01 mg/l
  • 0,01 mg/l
Specifications
Product Information
  • Catalog #
    Supplier #
    Type
    Pk
    Availability
    Price
  • FDO 700 IQ Optical, digital DO sensor suitable for water applications, without SACIQ cable
    FDO 700 IQ Optical, digital DO sensor suitable for water applications, without SACIQ cable
    706-0524
    201650
    FDO 700 IQ Optical
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 12 190 zł
    12 190 zł
    1 * 1 SZT
       12 190 zł
    Supplier Article Number: 201650
    Local Article Number: WTWO201650
    Specifications:
    More
    Accessories
    Specifications
    Model
    FDO 700 IQ
    Environmentally Preferable
    Min. flow rate
    No flow required
    Response time
    <150 s
    DO concentration
    0...20,00 mg/l
    DO saturation
    0...200,0%
    DO barometric pressure range
    10 bar
    DO accuracy
    ±0,05 mg/l
    DO resolution
    0,01 mg/l
  • FDO 700 IQ SW Optical, digital DO sensor specially suitable for salt water applications, without SACIQ cable
    FDO 700 IQ SW Optical, digital DO sensor specially suitable for salt water applications, without SACIQ cable
    706-0525
    201652
    FDO 700 IQ SW Optical
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 13 860 zł
    13 860 zł
    1 * 1 SZT
       13 860 zł
    Supplier Article Number: 201652
    Local Article Number: WTWO201652
    Specifications:
    More
    Accessories
    Specifications
    Model
    FDO 700 IQ SW
    Environmentally Preferable
    Min. flow rate
    No flow required
    Response time
    <60 s
    DO concentration
    0...20,00 mg/l
    DO saturation
    0...200,0%
    DO barometric pressure range
    10 bar
    DO accuracy
    ±0,05 mg/l
    DO resolution
    0,01 mg/l