Drukuj
Wyszukano: Stoły
38  wyniki/ów wyszukiwania
SearchResultCount:"38"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Collection
Opis: Granit polerowany. Cztery stabilne nóżki; pochłania i tłumi zewnętrzne wibracje, dzięki czemu nadaje się zarówno do precyzyjnego ważenia, jak również ...
Numer katalogowy: (611-2549)
Producent: VWR Collection
Opis: High performance micro balances and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast r...
j.m.: 1 * 1 SZT
Producent: Metro International
Opis: These stainless steel laboratory workbenches are available in two versions, with or without a bottom shelf. The versions without a bottom shelf have t...
Wyprzedaż
Producent: Palbam
Opis: These cleanroom heavy duty tables are designed for more robust and heavy duty activities and are manufactured to the highest cleanroom standards. The ...
Producent: Palbam
Opis: These cleanroom tables are manufactured to the highest cleanroom standards. The bases are fabricated from tubing for easy cleaning and optimal airflow...
Producent: Metro International
Opis: Cleanroom table in stainless steel 304 with electropolished finish and anodised satin connectors.
Producent: Sartorius Balances
Opis: High performance micro and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast stabilisat...
Producent: Palbam
Opis: The mobile laptop workstation provides a minimum footprint, stable work platform for working with laptops within the cleanroom.
Producent: Metro International
Opis: These stainless steel laboratory workbenches available top materials include type 304 stainless and TRESPA phenolic resin.
Producent: KOETTERMANN
Opis: Heavy duty mobile tables available with melamine TopResist® or laboratory grade SGL worktops.
Tables with both worktops suitable for:...
Producent: Palbam
Opis: The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
Producent: KOETTERMANN
Opis: These balance benches are designed to maintain vibration free condition, so that even small quantities can be weighed accurately.
Made from epoxy...
Producent: Palbam
Opis: These cleanroom ergo tables are manufactured with 304 stainless steel with electropolished finish and Al anodized cleanroom compatible connectors.
Producent: LABORIAL
Opis: Innovative interactive worktop for laboratories, cleanrooms and healthcare. A hygienic design based on GMP guidlines which consists of a continuous, t...
Producent: HUPFER METALLWERKE
Opis: This open version work table features closed table top and bottom side covered with 0,8 mm sheet steel.
Numer katalogowy: (HUPF7500184)
Producent: HUPFER METALLWERKE
Opis: This packing table is made up of high-quality stainless steel (both top and bottom). It can also be also as a work bench to pack surgical instruments.
j.m.: 1 * 1 SZT
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 38
no targeter for Bottom