Drukuj
Wyszukano: Generatory gazu
38  wyniki/ów wyszukiwania
SearchResultCount:"38"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Numer katalogowy: (CCDL928276)
Producent: LUCIDITY
Opis: Generator that produces all necessary gasses required to run the Lucidity GC-FID.
j.m.: 1 * 1 SZT
New Product
Numer katalogowy: (PEAK10-1295)
Producent: PEAK SCIENTIFIC
Opis: The NG5000(A) delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for...
j.m.: 1 * 1 SZT
Numer katalogowy: (PEAK10-2320)
Producent: PEAK SCIENTIFIC
Opis: The NG5000 delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC...
j.m.: 1 * 1 SZT
Numer katalogowy: (PEAK3300671)
Producent: PEAK SCIENTIFIC
Opis: Solaris XE 35 generator has been engineered using membrane technology to meet the gas delivery requirements for labs using LC-MS or multiple lower flo...
j.m.: 1 * 1 SZT
Numer katalogowy: (564-0277)
Producent: PEAK SCIENTIFIC
Opis: This nitrogen generator has been engineered and designed as a gas delivery solution for ELSD, Compact Mass Spectrometer instruments and TLD readers wh...
j.m.: 1 * 1 SZT
Numer katalogowy: (PEAK60-0030)
Producent: PEAK SCIENTIFIC
Opis: The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC at both ...
j.m.: 1 * 1 SZT
Producent: PEAK SCIENTIFIC
Opis: The smallest hydrogen generator for GC-FID, Precision hydrogen SL has been developed to offer a streamlined laboratory-grade hydrogen gas solution for...
Producent: PEAK SCIENTIFIC
Opis: Precision Hydrogen series hydrogen generators are suitable for carrier gas and flame gas at standard detection limits. Hydrogen purity: 99,9995%.
Numer katalogowy: (PEAK3300253)
Producent: PEAK SCIENTIFIC
Opis: Genius XE Nitrogen is a cutting-edge evolution combining advanced technology with refined and robust engineering. Genius XE Nitrogen provides a premiu...
j.m.: 1 * 1 SZT
Numer katalogowy: (564-0298)
Producent: PEAK SCIENTIFIC
Opis: The Precision Nitrogen Headspace 250 has been conceived and custom-engineered to meet the gas requirements for headspace sampler instruments requiring...
j.m.: 1 * 1 SZT
Numer katalogowy: (564-0280)
Producent: PEAK SCIENTIFIC
Opis: Designed to the specifications of the Bruker EVOQ triple Quadrupole's gas requirements delivering dry air at up to 50 L/min and nitrogen at up to 32 L...
j.m.: 1 * 1 SZT
Numer katalogowy: (564-0283)
Producent: PEAK SCIENTIFIC
Opis: Designed and engineered as an on-site, compressor less nitrogen generation solution tailored for the Shimadzu LCMS-8045, 8050 or 8060. The Infinity 10...
j.m.: 1 * 1 SZT
Numer katalogowy: (PEAK60-0018)
Producent: PEAK SCIENTIFIC
Opis: The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC at both ...
j.m.: 1 * 1 SZT
Producent: PEAK SCIENTIFIC
Opis: These precision nitrogen trace generators produce "zero nitrogen" on demand from a compressed air source.
Numer katalogowy: (PEAK109524)
Producent: PEAK SCIENTIFIC
Opis: The Genius 1024 nitrogen generator is designed to provide nitrogen flow to LCMS applications at a higher pressure.
j.m.: 1 * 1 SZT
Numer katalogowy: (PEAK81-4000)
Producent: PEAK SCIENTIFIC
Opis: The Corona 1010 meet the specifications of the Thermo Scientific Corona Veo Charged Aerosol Detector. This system can be purchased as a single nitroge...
j.m.: 1 * 1 SZT
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 38
no targeter for Bottom