Drukuj
Regulator mocy, MC227
Dostawca: Electrothermal
454-0041EA 2490 PLN
454-0041
Regulator mocy, MC227
Kontrolery Regulatory temperatury
This regulator controls the average energy input to a heating apparatus by automatically switching power on and off at the intervals regulated by the MC227. The temperature of the apparatus being regulated remains constant.
  • Suitable for bench, wall or 12.7 mm Ø scaffold rod mounting
Suitable for bench, wall, or scaffold rod mounting.
Maximum current intermittent: 10 A
Maximum continuous rating: 5 A
Dodatkowe informacje: Supplied without plug.