Drukuj
Dzielniki próbek, PT 100
Dostawca: Retsch
411-3311EA 54940 PLN
RETS40.535.0002
Dzielniki próbek, PT 100
Rotating sample divider PT100 for representative division/extraction of samples from the bulk. This procedure guarantees a very high degree of dividing accuracy and reproducibility with both fine and coarse materials.
  • Extremely high division accuracy
  • Representative and reproducible division for accurate analysis results
  • Automatic material feed via synchronized feeder
  • Simple and rapid handling due to quick-release clamping system for laboratory bottles
*depending on feed material and instrument configuration/settings.
Certyfikaty: IP Protection Class according to DIN EN 60529: IP 40.
Informacje do zamówienia: Complete PT100 unit includes dividing head with 8 quick-release outlets, DR 100/40 vibratory feeder and 10×250 ml wide-mouth bottles. Complete DR 100/40 unit includes feeding kit (40 mm push-fit feed chute, holder for 2,8 L push-fit chute hopper and clamping fixture).