Order Entry
Vacuum ovens, Vacutherm® series
undefined
Vacuum ovens, Vacutherm® series
  • Catalog No:
  • 466-2741
  • 466-2720
  • 466-0489
  • 466-2710
  • 466-2730
  • 466-0490
  • 466-1107
  • 466-2700
  • 466-2740
  • 466-1109
  • 466-1108
  • Model:
  • VT 6130 P-BL
  • VT 6130 M
  • VT 6025
  • VT 6060 M
  • VT 6060 P
  • VT 6025
  • VT 6060 M-BL
  • VT 6025
  • VT 6130 P
  • VT 6060 P-BL
  • VT 6130 M-BL
  • Temperature variation (spatial):
  • ±4 °C @ 200 °C
    ±7 °C @ 300 °C
  • ±6 °C @ 200 °C
  • ±4 °C @ 200 °C
  • ±4 °C @ 200 °C
  • ±3 °C @ 200 °C
    ±7 °C @ 300 °C
  • ±4 °C @ 200 °C
  • ±4 °C @ 200 °C
  • ±4 °C @ 200 °C
  • ±4 °C @ 200 °C
    ±7 °C @ 300 °C
  • ±3 °C @ 200 °C
    ±7 °C @ 300 °C
  • ±6 °C @ 200 °C
  • Temperature fluctuation (time):
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • ≤0,5 °C
  • Temperature range:
  • Ambient +10…300 °C
  • Ambient +15…200 °C
  • Ambient +15…200 °C
  • Ambient +15…200 °C
  • Ambient +10…300 °C
  • Ambient +15…200 °C
  • Ambient +15…200 °C
  • Ambient +15…200 °C
  • Ambient +10…300 °C
  • Ambient +10…300 °C
  • Ambient +15…200 °C
  • Capacity:
  • 128 L
  • 128 L
  • 25 L
  • 53 L
  • 53 L
  • 25 L
  • 53 L
  • 25 L
  • 128 L
  • 53 L
  • 128 L
  • Convection type:
  • Shelf heating
  • Air jacket system
  • Air jacket system
  • Air jacket system
  • Shelf heating
  • Air jacket system
  • Air jacket system
  • Air jacket system
  • Shelf heating
  • Shelf heating
  • Air jacket system
  • Ext. W×D×H:
  • 895×750×720 mm
  • 895×750×720 mm
  • 480×450×600 mm
  • 744×570×576 mm
  • 744×570×576 mm
  • 480×450×600 mm
  • 744×570×576 mm
  • 480×450×600 mm
  • 895×750×720 mm
  • 744×570×576 mm
  • 895×750×720 mm
  • Int. W×D×H:
  • 495×529×489** mm
  • 495×529×489 mm
  • 300×307×275 mm
  • 415×371×345 mm
  • 415×371×345* mm
  • 300×307×275 mm
  • 415×371×345 mm
  • 300×307×275 mm
  • 495×529×489** mm
  • 415×371×345* mm
  • 495×529×489 mm
  • Max. load per shelf:
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • 20 kg
  • Power consumption:
  • 3000 W
  • 2200 W
  • 1300 W
  • 1700 W
  • 1600 W
  • 1300 W
  • 1700 W
  • 1300 W
  • 3000 W
  • 1600 W
  • 2200 W
  • Shelves supplied (max.):
  • 3 (3)
  • 3 (5)
  • 2 (4)
  • 2 (4)
  • 2 (2)
  • 2 (4)
  • 2 (4)
  • 2 (4)
  • 3 (3)
  • 2 (2)
  • 3 (5)
  • Weight:
  • 164 kg
  • 153 kg
  • 58 kg
  • 82 kg
  • 90 kg
  • 58 kg
  • 82 kg
  • 58 kg
  • 164 kg
  • 90 kg
  • 153 kg
  •  
    Pk
  • VT 6025 with jacketed heating
    VT 6025 with jacketed heating
    466-2700
    51014550
    1
    1 * 1 SZT
    1 * 1 SZT - 40 720 zł
    40 720 zł
    1 * 1 SZT
       40 720 zł
    Accessories
    Specifications
    Model
    VT 6025
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    25 L
    Convection type
    Air jacket system
    Ext. W×D×H
    480×450×600 mm
    Int. W×D×H
    300×307×275 mm
    Max. load per shelf
    20 kg
    Power consumption
    1300 W
    Shelves supplied (max.)
    2 (4)
    Weight
    58 kg
  • VT 6025 with inert gas connection
    VT 6025 with inert gas connection
    466-0489
    51014552
    1
    1 * 1 SZT
    1 * 1 SZT - 46 350 zł
    46 350 zł
    1 * 1 SZT
       46 350 zł
    Accessories
    Specifications
    Model
    VT 6025
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    25 L
    Convection type
    Air jacket system
    Ext. W×D×H
    480×450×600 mm
    Int. W×D×H
    300×307×275 mm
    Max. load per shelf
    20 kg
    Power consumption
    1300 W
    Shelves supplied (max.)
    2 (4)
    Weight
    58 kg
  • VT 6025 with inert gas connection and digital pressure display
    VT 6025 with inert gas connection and digital pressure display
    466-0490
    51014553
    1
    1 * 1 SZT
    1 * 1 SZT - 67 400 zł
    67 400 zł
    1 * 1 SZT
       67 400 zł
    Accessories
    Specifications
    Model
    VT 6025
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    25 L
    Convection type
    Air jacket system
    Ext. W×D×H
    480×450×600 mm
    Int. W×D×H
    300×307×275 mm
    Max. load per shelf
    20 kg
    Power consumption
    1300 W
    Shelves supplied (max.)
    2 (4)
    Weight
    58 kg
  • VT 6060 M with jacketed heating
    VT 6060 M with jacketed heating
    466-2710
    51014539
    1
    1 * 1 SZT
    1 * 1 SZT - 65 730 zł
    65 730 zł
    1 * 1 SZT
       65 730 zł
    Accessories
    Specifications
    Model
    VT 6060 M
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    53 L
    Convection type
    Air jacket system
    Ext. W×D×H
    744×570×576 mm
    Int. W×D×H
    415×371×345 mm
    Max. load per shelf
    20 kg
    Power consumption
    1700 W
    Shelves supplied (max.)
    2 (4)
    Weight
    82 kg
  • VT 6060 M-BL with jacketed heating, for flammable solvents
    VT 6060 M-BL with jacketed heating, for flammable solvents
    466-1107
    51014546
    1
    1 * 1 SZT
    1 * 1 SZT - 95 810 zł
    95 810 zł
    1 * 1 SZT
       95 810 zł
    Accessories
    Specifications
    Model
    VT 6060 M-BL
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    53 L
    Convection type
    Air jacket system
    Ext. W×D×H
    744×570×576 mm
    Int. W×D×H
    415×371×345 mm
    Max. load per shelf
    20 kg
    Power consumption
    1700 W
    Shelves supplied (max.)
    2 (4)
    Weight
    82 kg
  • VT 6130 M with jacketed heating
    VT 6130 M with jacketed heating
    466-2720
    51014541
    1
    1 * 1 SZT
    1 * 1 SZT - 94 690 zł
    94 690 zł
    1 * 1 SZT
       94 690 zł
    Accessories
    Specifications
    Model
    VT 6130 M
    Environmentally Preferable
    Temperature variation (spatial)
    ±6 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    128 L
    Convection type
    Air jacket system
    Ext. W×D×H
    895×750×720 mm
    Int. W×D×H
    495×529×489 mm
    Max. load per shelf
    20 kg
    Power consumption
    2200 W
    Shelves supplied (max.)
    3 (5)
    Weight
    153 kg
  • VT 6130 M-BL with jacketed heating, for flammable solvents
    VT 6130 M-BL with jacketed heating, for flammable solvents
    466-1108
    51014547
    1
    1 * 1 SZT
    1 * 1 SZT - 121 430 zł
    121 430 zł
    1 * 1 SZT
       121 430 zł
    Accessories
    Specifications
    Model
    VT 6130 M-BL
    Environmentally Preferable
    Temperature variation (spatial)
    ±6 °C @ 200 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +15…200 °C
    Capacity
    128 L
    Convection type
    Air jacket system
    Ext. W×D×H
    895×750×720 mm
    Int. W×D×H
    495×529×489 mm
    Max. load per shelf
    20 kg
    Power consumption
    2200 W
    Shelves supplied (max.)
    3 (5)
    Weight
    153 kg
  • VT 6060 P with shelf heating
    VT 6060 P with shelf heating
    466-2730
    51014542
    1
    1 * 1 SZT
    1 * 1 SZT - 99 710 zł
    99 710 zł
    1 * 1 SZT
       99 710 zł
    Accessories
    Specifications
    Model
    VT 6060 P
    Environmentally Preferable
    Temperature variation (spatial)
    ±3 °C @ 200 °C
    ±7 °C @ 300 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +10…300 °C
    Capacity
    53 L
    Convection type
    Shelf heating
    Ext. W×D×H
    744×570×576 mm
    Int. W×D×H
    415×371×345* mm
    Max. load per shelf
    20 kg
    Power consumption
    1600 W
    Shelves supplied (max.)
    2 (2)
    Weight
    90 kg
  • VT 6060 P-BL with shelf heating, for flammable solvents
    VT 6060 P-BL with shelf heating, for flammable solvents
    466-1109
    51014548
    1
    1 * 1 SZT
    1 * 1 SZT - 132 010 zł
    132 010 zł
    1 * 1 SZT
       132 010 zł
    Accessories
    Specifications
    Model
    VT 6060 P-BL
    Environmentally Preferable
    Temperature variation (spatial)
    ±3 °C @ 200 °C
    ±7 °C @ 300 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +10…300 °C
    Capacity
    53 L
    Convection type
    Shelf heating
    Ext. W×D×H
    744×570×576 mm
    Int. W×D×H
    415×371×345* mm
    Max. load per shelf
    20 kg
    Power consumption
    1600 W
    Shelves supplied (max.)
    2 (2)
    Weight
    90 kg
  • VT 6130 P with shelf heating
    VT 6130 P with shelf heating
    466-2740
    51014543
    1
    1 * 1 SZT
    1 * 1 SZT - 150 390 zł
    150 390 zł
    1 * 1 SZT
       150 390 zł
    Accessories
    Specifications
    Model
    VT 6130 P
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    ±7 °C @ 300 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +10…300 °C
    Capacity
    128 L
    Convection type
    Shelf heating
    Ext. W×D×H
    895×750×720 mm
    Int. W×D×H
    495×529×489** mm
    Max. load per shelf
    20 kg
    Power consumption
    3000 W
    Shelves supplied (max.)
    3 (3)
    Weight
    164 kg
  • VT 6130 P-BL with shelf heating, for flammable solvents
    VT 6130 P-BL with shelf heating, for flammable solvents
    466-2741
    51014549
    1
    1 * 1 SZT
    1 * 1 SZT - 170 450 zł
    170 450 zł
    1 * 1 SZT
       170 450 zł
    Accessories
    Specifications
    Model
    VT 6130 P-BL
    Environmentally Preferable
    Temperature variation (spatial)
    ±4 °C @ 200 °C
    ±7 °C @ 300 °C
    Temperature fluctuation (time)
    ≤0,5 °C
    Temperature range
    Ambient +10…300 °C
    Capacity
    128 L
    Convection type
    Shelf heating
    Ext. W×D×H
    895×750×720 mm
    Int. W×D×H
    495×529×489** mm
    Max. load per shelf
    20 kg
    Power consumption
    3000 W
    Shelves supplied (max.)
    3 (3)
    Weight
    164 kg