Drukuj
Profesjonalne lodówki, serii GKv i GKPv
Dostawca: Liebherr
471-1015EA 5260 PLN
471-1015 LIEBGKV5790 LIEBGKV6410924
Profesjonalne lodówki, serii GKv i GKPv
Chłodziarki/Lodówki
These commercial refrigerators feature a fan for forced air cooling with temperature adjustable from –2 to +15 °C or 1 to 15 °C.
  • Electronic temperature regulation, with digital display
  • Alarm for temperature and door opening
  • Automatic defrosting with evaporation of condensates
  • Interior with rounded corners to make cleaning easier
  • Door is lockable and reversible, self closing at 45° but stable at 90°
ABS interior, exterior made from white epoxy coated steel or stainless steel, high density insulation 50 mm (allows temperature control even at +43 °C).
Dodatkowe informacje: These refrigerator cabinets are delivered with adjustable feet (150 to 180 mm).