Drukuj
Wyszukano: Dejonizatory
15  wyniki/ów wyszukiwania
SearchResultCount:"15"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Numer katalogowy: (171-2708)
Producent: VWR Collection
Opis: This demineraliser connects to a mains water tap and instantly delivers pure water with high resistivity. The exchange capacity of a cartridge is 300&...
j.m.: 1 * 1 SZT
Producent: MIELE
Opis: Aqua purifier for two full or partial demineralisation cartridges. The casing made from white steel or stainless steel.
Numer katalogowy: (141-0552)
Producent: MIELE
Opis: The Aqua-Soft system provides a continual supply of soft water and is suitable for water with a hardness up to 71° dH. It operates as a level controll...
j.m.: 1 * 1 SZT
Numer katalogowy: (BIND8012-0759)
Producent: Binder
Opis: This system is used to fully desalinate faucet water for BINDER climate chamber humidification systems. The ion exchanger system consists of a pressur...
j.m.: 1 * 1 SZT
Numer katalogowy: (172-6111)
Producent: EVOQUA WATER TECHNOLOGIES GMBH
Opis: Continuous electrodeionisation is a safe, chemical-free way to take RO (reverse osmosis) water to a higher level of purity. This water deionisation un...
j.m.: 1 * 1 SZT
Producent: HYDROLAB
Opis: Demineralizatory serii BASIC to urządzenia do produkcji wody zdemineralizowanej wprost z wody wodociągowej. W pełni automatyczne i bezobsługowe urządz...
Numer katalogowy: (171-0484)
Producent: ELGA LabWater
Opis: This wall mounted disposable deioniser produces water up to a quality of 10M.-cm at flows of up to 90 l/hr. Designed for users with a requirement for ...
j.m.: 1 * 1 SZT
Producent: ENVIROFALK PHARMAWATERSYSTEMS
Opis: Pressure-resistant water deionisation cartridges made from stainless steel, filled with regenerable mixed-bed resins.
Producent: BARNSTEAD (INC THERMOLYNE)
Opis: These B-Pure™ laboratory-grade water systems are designed for pre-treatment or deionization application needs. The design provides the ability to add ...
Numer katalogowy: (HECH40075010)
Producent: GLASWARENFABRIK KARL HECHT
Opis: Ion exchanger for purified water.
j.m.: 1 * 1 SZT
Numer katalogowy: (BARND0805)
Producent: BARNSTEAD (INC THERMOLYNE)
Opis: It is a simple, cost effective method for deionisation or pretreatment of lab water. Typically these units are ideal where water usage is up to 38 lit...
j.m.: 1 * 1 SZT
Producent: BEHR
Opis: Mixed bed ion exchanger for small to medium quantities of ultrapure water to the supply from laboratory dishwashers to general care in the laboratory ...
Producent: ENVIROFALK PHARMAWATERSYSTEMS
Opis: Pressureless water deionisation cartridge with plastic housing, filled with regenerable mixed-bed resins.
Producent: BEHR
Opis: These deionisers are thick-walled, robust and practical mixed-bed ion exchangers made of blue polyethylene with free output to the storage tank. Ideal...
Numer katalogowy: (BEHRB00223482)
Producent: BEHR
Opis: The standard canister is made of V4A stainless steel with widespread pure water supply applications. They are aslo suitable for second stage purificat...
j.m.: 1 * 1 SZT
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 15 of 15
no targeter for Bottom