Order Entry
Reverse osmosis systems, ReversinO
Reverse osmosis systems, ReversinO
Certificates
undefined
Reverse osmosis systems, ReversinO
Specifications
  • Catalog No:
  • 172-4216
  • 172-4215
  • Model:
  • ReversinO 6 Plus
  • ReversinO 3 Plus
  • Feed water:
  • Tap water
  • Tap water
  • Production flow rate:
  • 4 - 12 L/h
  • 2 - 6 L/h
  • W×D×H:
  • 370×250×700 mm
  • 370×250×700 mm
  • Weight:
  • 12 kg
  • 11,5 kg
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • ReversinO 6 Plus
    ReversinO 6 Plus
    172-4216
    1981-041
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 1981-041
    Local Article Number: WERN1981-041
    Specifications:
    More
    Specifications
    Model
    ReversinO 6 Plus
    Environmentally Preferable
    Feed water
    Tap water
    Production flow rate
    4 - 12 L/h
    W×D×H
    370×250×700 mm
    Weight
    12 kg
  • ReversinO 3 Plus
    ReversinO 3 Plus
    172-4215
    1981-040
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 1981-040
    Local Article Number: WERN1981-040
    Specifications:
    More
    Specifications
    Model
    ReversinO 3 Plus
    Environmentally Preferable
    Feed water
    Tap water
    Production flow rate
    2 - 6 L/h
    W×D×H
    370×250×700 mm
    Weight
    11,5 kg