The Elmasonic P ultrasonic series is ideal for all applications in analytical and medical laboratories and in industrial production. Six different sizes are available. These practical units are easy to operate with a clearly arranged display showing all set and actual parameters. Several special features make the units perfect for intensive cleaning applications. All models are equipped with a mechanical timer up to maximum of 15 minutes.
- Clear display
- Easy and intuitive setting
- Power-controlled ultrasound with high energy density for difficult applications (low frequency) or gentle and quiet for the cleaning of capillaries and small items (high frequency)
- The temperature can be set from 30 to 80 °C in increments of 5 °C
Multi-frequency at 37 kHz; standard applications are carried out at 37 kHz. All units are considerably more powerful than conventional ultrasonic units. If the high energy density should prove too stong for individual cleaning jobs, it can be reduced electronically.
Multi-frequency at 80 kHz; operating the unit at 80 kHz prolongs the cleaning time and is ideal for the treatment of very small and sensitive items. Even tiny capillaries are reached by the imploding microscopic cavitation bubbles. A positive side effect is the comfortably low noise at this frequency.
Special ultrasonic features:
Standard: For applications in the laboratory such as mixing, dissolving, dispersing
Pulse: Activatable 'Power' addition of ~20% through increased peak performance
Sweep: For an even distribution of the ultrasonic power in the cleaning bath
Degas: For quick degassing of samples or solvents
Power regulation: Stepwise reduction of the ultrasonic power for the cleaning of sensitive surfaces
Two additional features are available:
Pause: Interrupt and restart the current application
Temperature control: Automatic start-up of the ultrasound when the set temperature is reached
- Catalog No:
- 142-0050
- 142-0049
- 142-0051
- 142-0047
- 142-0048
- 142-0046
- Model:
- P 300 H
- P 180 H
- P 30 H
- P 60 H
- P 120 H
- P 70 H
- Capacity:
- 28,0 L
- 18,0 L
- 2,75 L
- 5,75 L
- 12,75 L
- 6,9 L
- Frequency:
- 37 / 80 kHz
- 37 / 80 kHz
- 37 / 80 kHz
- 37 / 80 kHz
- 37 / 80 kHz
- 37 / 80 kHz
- Heating power:
- 1200 W
- 800 W
- 200 W
- 400 W
- 800 W
- 600 W
- Power consumption:
- 1580 W
- 1130 W
- 320/300 W
- 580/550 W
- 1130 W
- 820 W
- Tank W×D×H:
- 505×200×300 mm
- 327×200×300 mm
- 240×100×137 mm
- 300×150×151 mm
- 300×200×240 mm
- 505×100×137 mm
- W×D×H:
- 568×321×340 mm
- 390×321×340 mm
- 300×221×179 mm
- 365×271×186 mm
- 365×321×278 mm
- 568×221×179 mm
- Pk
- Ultrasonic bath P 30 H
Accessories
Specifications
ModelP 30 HCapacity2,75 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power200 WPower consumption320/300 WTank W×D×H240×100×137 mmW×D×H300×221×179 mm - Ultrasonic bath P 60 H
Accessories
Specifications
ModelP 60 HCapacity5,75 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power400 WPower consumption580/550 WTank W×D×H300×150×151 mmW×D×H365×271×186 mm - Ultrasonic bath P 70 H
Accessories
Specifications
ModelP 70 HCapacity6,9 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power600 WPower consumption820 WTank W×D×H505×100×137 mmW×D×H568×221×179 mm - Ultrasonic bath P 120 HUltrasonic bath P 120 H142-0048101377711 * 1 SZT11 830 zł
Accessories
Specifications
ModelP 120 HCapacity12,75 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power800 WPower consumption1130 WTank W×D×H300×200×240 mmW×D×H365×321×278 mm - Ultrasonic bath P 180 HUltrasonic bath P 180 H142-0049101380411 * 1 SZT13 020 zł
Accessories
Specifications
ModelP 180 HCapacity18,0 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power800 WPower consumption1130 WTank W×D×H327×200×300 mmW×D×H390×321×340 mm - Ultrasonic bath P 300 HUltrasonic bath P 300 H142-0050101381011 * 1 SZT15 130 zł
Accessories
Specifications
ModelP 300 HCapacity28,0 LEnvironmentally PreferableFrequency37 / 80 kHzHeating power1200 WPower consumption1580 WTank W×D×H505×200×300 mmW×D×H568×321×340 mm
Baskets
- Pk
Covers
- SeqDescriptionForPkPrice
Modular Basket Systems
- SeqDescriptionPk
- 06Modular basket system type 80, chamfer 20×20 mm, dimension: 455×105×55 mm1