Drukuj
Pojemniki na ostre odpady zakaźne, Sharpsafe®, HUART-HOSPIDEX (MSH)
Dostawca: Hospidex
Sharpsafe® Classics
129-1809EA 3120 PLN
129-1809 129-1192 129-1190 119-0089 129-1191 119-0087 119-0086 119-0085 HUARSHA-40502610 119-0084 119-0082 119-0081 119-0080 129-1181 119-0079 129-1182 119-0077 129-1180 119-0076 129-1189 129-1187 129-1188 129-1185 129-1186 129-1183 129-1184
Pojemniki na ostre odpady zakaźne, Sharpsafe®, HUART-HOSPIDEX (MSH)
Kosze na odpady Kosze na odpady ostre
PP.
  • Disconnection of all needles with Luer lock and Luer slip connection (except 119-0089, -0086, 129-1809)
  • Suitable for the removal of pen injector insulin needles (except 129-1809, 119-0086, -0089, -0087)
  • Disconnection of scalpel blades (except 119-0089, -0086, 129-1809, 119-0084)
TRBA 250, NF X30-500, NF 302 and UN certified
Colour: Yellow. Translucent cover.