Drukuj
Fotometry z filtrem, photoLab® S12
Dostawca: WTW
photoLab®
713-3004EA 18220 PLN
713-3004 713-3003
Fotometry z filtrem, photoLab® S12
Spektrofotometry i fotometry Spektrofotometry UV/Vis
Filter photometer with 12 wavelengths for extensive use in water analysis.
  • Large number of samples especially in the water testing industry
  • Fast accurate results, barcode test identification, when used in conjunction with WTW and Merck test kits
  • 50 user-defined methods are possible and reaction kinetics can be monitored
Certyfikaty: AQA/IQC multi-level