Dexamethasone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-02-2
:
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136
:392.47 g/mol
Melting Pt:268…271 °C
:M-Clarity™ quality level = MQ200
EINECS:200-003-9
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: D4902-25MGLocal Article Number: SIALD4902-25MG
- Supplier Article Number: D4902-100MGLocal Article Number: SIALD4902-100MG
- SIALD4902-500MGD4902-500MGGlass bottle for solids500 mgLog in for availability1 * 500 mg2 120 złSupplier Article Number: D4902-500MGLocal Article Number: SIALD4902-500MG
- Supplier Article Number: D4902-1GLocal Article Number: SIALD4902-1G