Drukuj
Lejki bezpieczeństwa z pokrywką na zawiasach, SCAT EUROPE
Dostawca: SCAT Europe
590-0033EA 1320 PLN
590-0033 221-0212
Lejki bezpieczeństwa z pokrywką na zawiasach, SCAT EUROPE
Lejki, rozdzielacze i wkraplacze Lejki bezpieczeństwa
HDPE, white or HDPE, black, electroconductive with grounding gripper.
  • Removable sieves for cleaning
  • Low installed height, even fits below low work surfaces
  • Various thread diameters
Suitable for use with with most commonly used chemicals. The screw cap rotates freely, which makes it easier to screw the funnel on and off.