Membrane filters, type 154
Supplier: Sartorius
New Product
Certificates
About this item
PES membrane filters are hydrophilic, have high flow rates thanks to the symmetric structure, have a low non-specific protein adsorption and are chemically resistant over a pH range of 1 to 14. They are recommended for the filtration of aqueous solutions as well for the protein filtration. Furthermore, the low level of extractables makes them suitable for environmental analysis.
Specifications
- Catalog No:
- SART15407-50-MIN
- SART15406-50-N
- 512-5076
- 512-2954
- SART15407-47-MIN
- SART15407-25-MIN
- SART15458-47-N
- SART15406-25-N
- SART15407-142-MIG
- SART15406-47-N
- Type:
- 154
- 154
- 154
- 154
- 154
- 154
- 154
- 154
- 154
- 154
- Pore size:
- 0,20 µm
- 0,45 µm
- 0,10 µm
- 0,10 µm
- 0,20 µm
- 0,20 µm
- 0,10 µm
- 0,45 µm
- 0,20 µm
- 0,45 µm
- Thickness:
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- 150 μm
- Water flow rate:
- >28 ml/min/cm² (1 bar)
- >32 ml/min/cm² (1 bar)
- >7 ml/min/cm² (1 bar)
- >7 ml/min/cm² (1 bar)
- >28 ml/min/cm² (1 bar)
- >28 ml/min/cm² (1 bar)
- >7 ml/min/cm² (1 bar)
- >32 ml/min/cm² (1 bar)
- >28 ml/min/cm² (1 bar)
- >32 ml/min/cm² (1 bar)
Specifications
Product Information
- Catalog #Supplier #TypePore sizeØPkAvailabilityPrice
- Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,10 µmThickness150 μmWater flow rate>7 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,10 µmThickness150 μmWater flow rate>7 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,10 µmThickness150 μmWater flow rate>7 ml/min/cm² (1 bar) - Specifications:New Product
More
Specifications
Type154Environmentally PreferablePore size0,20 µmThickness150 μmWater flow rate>28 ml/min/cm² (1 bar)New Product - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,20 µmThickness150 μmWater flow rate>28 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,20 µmThickness150 μmWater flow rate>28 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,20 µmThickness150 μmWater flow rate>28 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,45 µmThickness150 μmWater flow rate>32 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,45 µmThickness150 μmWater flow rate>32 ml/min/cm² (1 bar) - Specifications:
More
Specifications
Type154Environmentally PreferablePore size0,45 µmThickness150 μmWater flow rate>32 ml/min/cm² (1 bar)