trans-2-Nonenal 95%
ACRO298310250
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4520 to 1.4560 (20°C, 589 nm)
- GC:≥94 %
: (E)-2-Nonenal
:
:140.23 g/mol
:12,06771
:98…100 °C (12 mmHg)
:0.846 g/cm³
:84 °C
:242-609-6
:GHS07
:Refrigerator
:18829-56-6
:C₉H₁₆O
:MFCD00007012