Water purification system, Milli-Q® Integral
: Merck
About this item
The Milli-Q® Integral system is a compact solution that combines the production of Type II (pure) water and Type I (ultrapure) water in a single unit, eliminating the need for a pretreatment stage upstream of the ultrapure water system.
- Integrated Elix® technology provides high, consistent water quality at optimum operating cost
- Resistivity and TOC monitoring allows control over both ionic and organic contaminants that can impact results
- Placed at the outlet of each dispenser, the system's POD Paks have been designed to remove specific types of contaminants
- Catalog No:
- 171-0661
- 171-0662
- 171-0716
- 171-0725
- 171-0734
- 171-0743
- Model:
- Milli-Q® Integral 5
- Milli-Q® Integral 10
- Milli-Q® Integral 3
- Milli-Q® Integral 5
- Milli-Q® Integral 10
- Milli-Q® Integral 15
- Flow rate:
- Up to 2,0 l/min (at the E-POD® level)
- Up to 2,0 l/min (at the E-POD® level)
- Up to 2,0 l/min (at the E-POD® level)
- Up to 2,0 l/min (at the E-POD® level)
- Up to 2,0 l/min (at the E-POD® level)
- Up to 2,0 l/min (at the E-POD® level)
- Production flow rate:
- 5 L/h
- 10 L/h
- 3 L/h
- 5 L/h
- 10 L/h
- 15 L/h
- TOC content:
- <5 ppb
- <5 ppb
- <5 ppb
- <5 ppb
- <5 ppb
- <5 ppb
- Resistivity at +25 °C:
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- Bacteria:
- <0,1
- <0,1
- <0,1
- <0,1
- <0,1
- <0,1
- Particles:
- <1
- <1
- <1
- <1
- <1
- <1
- Pyrogen:
- < 0,001 (with BioPak filter as POD Pak)
- < 0,001 (with BioPak filter as POD Pak)
- < 0,001 (with BioPak filter as POD Pak)
- < 0,001 (with BioPak filter as POD Pak)
- < 0,001 (with BioPak filter as POD Pak)
- < 0,001 (with BioPak filter as POD Pak)
- RNases:
- <0,01
- <0,01
- <0,01
- <0,01
- <0,01
- <0,01
- DNases:
- <4
- <4
- <4
- <4
- <4
- <4
- W×D×H:
- 332×484×500 mm
- 332×484×500 mm
- 332×484×500 mm
- 332×484×500 mm
- 332×484×500 mm
- 332×484×500 mm
- Operating weight with full tank:
- 24…28 kg
- 24…28 kg
- 24…28 kg
- 24…28 kg
- 24…28 kg
- 24…28 kg
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Water purification system, Milli-Q® Integral 5 (EU-plug, manual in English)Water purification system, Milli-Q® Integral 5 (EU-plug, manual in English)171-0661ZRXQ005EU11 * 1 SZT96 570 zł
Accessories
Specifications
ModelMilli-Q® Integral 5Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate5 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg - Water purification system, Milli-Q® Integral 10 (EU-plug, manual in English)Water purification system, Milli-Q® Integral 10 (EU-plug, manual in English)171-0662ZRXQ010EU11 * 1 SZT113 640 zł
Accessories
Specifications
ModelMilli-Q® Integral 10Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate10 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg - Water purification system, Milli-Q® Integral 3Water purification system, Milli-Q® Integral 3171-0716ZRXQ003FRSAVT11 * 1 SZT
Accessories
Specifications
ModelMilli-Q® Integral 3Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate3 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg - Water purification system, Milli-Q® Integral 5Water purification system, Milli-Q® Integral 5171-0725ZRXQ005FRSAVT11 * 1 SZT
Accessories
Specifications
ModelMilli-Q® Integral 5Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate5 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg - Water purification system, Milli-Q® Integral 5Water purification system, Milli-Q® Integral 5171-0734ZRXQ010FRSAVT11 * 1 SZT
Accessories
Specifications
ModelMilli-Q® Integral 10Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate10 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg - Water purification system, Milli-Q® Integral 15Water purification system, Milli-Q® Integral 15171-0743ZRXQ015FRSAVT11 * 1 SZT
Accessories
Specifications
ModelMilli-Q® Integral 15Environmentally PreferableFlow rateUp to 2,0 l/min (at the E-POD® level)Production flow rate15 L/hTOC content<5 ppbResistivity at +25 °C18,2 MΩ.cmBacteria<0,1Particles<1Pyrogen< 0,001 (with BioPak filter as POD Pak)RNases<0,01DNases<4W×D×H332×484×500 mmOperating weight with full tank24…28 kg
Cartridges
- SeqDescriptionPkPrice