Order Entry
Graduated pipettes, type 1, Class AS, works certified, MBL
Graduated pipettes, type 1, Class AS, works certified, MBL
Supplier:  SciLabware
Certificates
undefined
Graduated pipettes, type 1, Class AS, works certified, MBL
Supplier:  SciLabware
Product Information
  • Catalog #
    Supplier #
    Capacity
    Division
    Tolerance
    Colour code
    Pk
    Availability
    Price
  • 612-3949
    PRA788ASWC
    25 ml
    0,20 ml
    0,100 ± ml
    Green
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 184 zł
    184 zł
    1 * 2 SZT
       184 zł
    Supplier Article Number: PRA788ASWC
    Local Article Number: BIBBPRA788ASWC
    Specifications:
    Specifications
    Environmentally Preferable
  • 612-3948
    PRA786ASWC
    10 ml
    0,10 ml
    0,050 ± ml
    Orange
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 130 zł
    130 zł
    1 * 2 SZT
       130 zł
    Supplier Article Number: PRA786ASWC
    Local Article Number: BIBBPRA786ASWC
    Specifications:
    Specifications
    Environmentally Preferable
  • 612-3945
    PRA780ASWC
    1 ml
    0,01 ml
    0,007 ± ml
    Yellow
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 125 zł
    125 zł
    1 * 2 SZT
       125 zł
    Supplier Article Number: PRA780ASWC
    Local Article Number: BIBBPRA780ASWC
    Specifications:
    Specifications
    Environmentally Preferable
  • 612-3947
    PRA784ASWC
    5 ml
    0,05 ml
    0,030 ± ml
    Red
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 127 zł
    127 zł
    1 * 2 SZT
       127 zł
    Supplier Article Number: PRA784ASWC
    Local Article Number: BIBBPRA784ASWC
    Specifications:
    Specifications
    Environmentally Preferable
  • 612-3946
    PRA782ASWC
    2 ml
    0,02 ml
    0,010 ± ml
    Black
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 126 zł
    126 zł
    1 * 2 SZT
       126 zł
    Supplier Article Number: PRA782ASWC
    Local Article Number: BIBBPRA782ASWC
    Specifications:
    Specifications
    Environmentally Preferable