Barium sulphide, Sigma-Aldrich®
SIAL523437-10G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:12,01024
:1999.85 °C
:4.25 g/cm³ (20 °C)
:4.3,III
:169.4 g/mol
:244-214-4
:3134
:GHS07
:GHS09
:21109-95-5
:BaS
:MFCD00014192