Poland
Order Entry
Barium sulphide, Sigma-Aldrich®
Barium sulphide, Sigma-Aldrich®
  SIAL523437-10G
 :  Merck
 :  
Barium sulphide, Sigma-Aldrich®
  SIAL523437-10G
 :  Merck
 :  
622 zł
  1 * 10 g
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236
 :
 :12,01024
 :1999.85 °C
 :4.25 g/cm³ (20 °C)
 :4.3,III
 :169.4 g/mol
 :244-214-4
 :3134
 :GHS07
 :GHS09
 :21109-95-5
 :BaS
 :MFCD00014192