Drukuj

2-Nitro-4-(trifluoromethyl)benzonitrile, Sigma-Aldrich®

Dostawca: Merck


M-Clarity™ quality level = MQ100

SIAL329835-5GEA 637 PLN
SIAL329835-5G
2-Nitro-4-(trifluoromethyl)benzonitrile, Sigma-Aldrich®
2-Nitro-4-(trifluoromethyl)benzonitrile
Formuła: C₈H₃F₃N₂O₂
MW: 216,12 g/mol
Numer MDL: MFCD00014684
Numer CAS: 778-94-9
Numer WE (dawny EINECS): 212-298-1
Jednostka: 3439
ADR: 6.1,III

Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR