

Benzene ≥99%
numer katalogowy L14012.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Supplier Article Number: L14012.AP
Local Article Number: ALFAL14012.AP
Discontinued
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
Specifications
About this item
:
:78.11 g/mol
Merck Index:14,1066
Boiling Pt:80.1 °C (1013 hPa)
Melting Pt:5.5 °C
Density:0.878 g/cm³ (20 °C)
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:200-753-7
UN:1114
ADR:3,II
REACH:01-2119447106-44
:GHS02
:GHS08
CAS Number:71-43-2
Formula:C₆H₆
MDL Number:MFCD00003009