Order Entry
Bunsen burners, with stopcock
Bunsen burners, with stopcock
Supplier:  BOCHEM
Certificates
undefined
Bunsen burners, with stopcock
Supplier:  BOCHEM
Product Information
  • Catalog #
    Supplier #
    For
    Max. temp.
    Pk
    Availability
    Price
  • Bunsen burner with stopcock, pilot flame and air regulation
    Bunsen burner with stopcock, pilot flame and air regulation
    BOCH7050
    7050
    Natural gas
    1300 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 070 zł
    1 070 zł
    1 * 1 SZT
       1 070 zł
    Supplier Article Number: 7050
    Local Article Number: BOCH7050
    Specifications:
    Specifications
    Environmentally Preferable
  • Bunsen burner with stopcock, pilot flame and air regulation
    Bunsen burner with stopcock, pilot flame and air regulation
    BOCH7060
    7060
    Propane gas
    1300 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 070 zł
    1 070 zł
    1 * 1 SZT
       1 070 zł
    Supplier Article Number: 7060
    Local Article Number: BOCH7060
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together