Culture tubes, sterile
Dostawca: Simport Scientific
Certyfikaty
Informacje o tym produkcie
Made of polystyrene, transparent or polypropylene, translucent.
- Max. RCF: 3000 xg (PP); 1400 xg (PS)
- Resist temperatures from -190 °C to +120 °C (PP) and up to 70 °C (PS)
These disposable sterile tubes can be used for most routine laboratory procedures. They are biologically inert and exempt from mold release agents. Precision molding with virgin thermoplastics ensures that the tubes will be uniform in size and shape. High resistance to breakage reduces danger in handling infectious or other potentially harmful cultures.
Tubes are supplied with either a 2-position ribbed polyethylene cap (which can be left loose for aerobic work or sealed for anaerobic cultures) or without caps for general purpose work.
Tubes, PS
Informacje o produkcie
- Catalog #Supplier #CapacityPackedHeightØPkDostępnośćCena
- With capWith capSIMPT415-2T415-25 ml25 per bag/500 per case75 mm12 mm500Zaloguj się dla dostępności1 * 500 SZT905 złSupplier Article Number: T415-2Local Article Number: SIMPT415-2Dane techniczne:
Specifications
Environmentally Preferable - Without capWithout capSIMPT416-6T416-614 ml125 per bag/1000 per case95 mm17 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 520 złSupplier Article Number: T416-6Local Article Number: SIMPT416-6Dane techniczne:
Specifications
Environmentally Preferable - With capWith capSIMPT415-3T415-35 ml125 per bag/1000 per case75 mm12 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 330 złSupplier Article Number: T415-3Local Article Number: SIMPT415-3Dane techniczne:
Specifications
Environmentally Preferable - Without capWithout capSIMPT415-6T415-65 ml125 per bag/1000 per case75 mm12 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 260 złSupplier Article Number: T415-6Local Article Number: SIMPT415-6Dane techniczne:
Specifications
Environmentally Preferable - With capWith capSIMPT416-2T416-214 ml25 per bag/500 per case95 mm17 mm500Zaloguj się dla dostępności1 * 500 SZT1 200 złSupplier Article Number: T416-2Local Article Number: SIMPT416-2Dane techniczne:
Specifications
Environmentally Preferable - With capWith capSIMPT416-3T416-314 ml125 per bag/1000 per case95 mm17 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 590 złSupplier Article Number: T416-3Local Article Number: SIMPT416-3Dane techniczne:
Specifications
Environmentally Preferable
Tubes, PP
Informacje o produkcie
- Catalog #Supplier #CapacityPackedHeightØPkDostępnośćCena
- With capWith capSIMPT415-2AT415-2A5 ml25 per bag/500 per case75 mm12 mm500Zaloguj się dla dostępności1 * 500 SZT960 złSupplier Article Number: T415-2ALocal Article Number: SIMPT415-2ADane techniczne:
Specifications
Environmentally Preferable - Without capWithout capSIMPT416-6AT416-6A14 ml125 per bag/1000 per case95 mm17 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 780 złSupplier Article Number: T416-6ALocal Article Number: SIMPT416-6ADane techniczne:
Specifications
Environmentally Preferable - Without capWithout capSIMPT415-6AT415-6A5 ml125 per bag/1000 per case75 mm12 mm1.000Zaloguj się dla dostępności1 * 1.000 SZT1 330 złSupplier Article Number: T415-6ALocal Article Number: SIMPT415-6ADane techniczne:
Specifications
Environmentally Preferable - With capWith capSIMPT416-2AT416-2A14 ml25 per bag/500 per case95 mm17 mm500Zaloguj się dla dostępności1 * 500 SZT935 złSupplier Article Number: T416-2ALocal Article Number: SIMPT416-2ADane techniczne:
Specifications
Environmentally Preferable