Drukuj
Lancety do krwi
Dostawca: Marienfeld
MARI7300200EA 20.2 PLN
MARI7300200
Lancety do krwi
Lancety do krwi
Made of stainless steel of high grade quality X6 Cr 17 (in compliance with DIN EN 10088-2), chrome content 16 to 18%.
  • Corrosion-resistant, free from nickel
  • With tip optimized for painless blood taking
  • Sterilized by gamma irradiation in compliance with DIN EN 556
  • For single use
Sposób pakowania: Individually packed in paper, 5 pieces in a stripe; 200 pieces in a box; 5 boxes in a carton, 10 cartons in an outer carton.